Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 21162229-21162500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIZ
Entrez Gene ID
55857
Gene Name
kizuna centrosomal protein
Gene Aliases
C20orf19, HT013, Kizuna, NCRNA00153, PLK1S1, RP69
UniGene
Hs.187635
Gene Chromosome Location
Chr.20: 21125980-21246628 on Build GRCh38
Transcripts
NM_001163022.1 NM_001163023.1 NM_001276389.1 NM_018474.4 XM_011529296.2 XM_011529297.2 XM_011529298.2 XM_011529299.2 XM_017027951.1 XM_017027952.1 XM_017027953.1 XM_017027954.1

SNP Detail

SNP ID Chromosome Location
rs780070529 Chr.20: 21162296-21162296 on Build GRCh38
rs181816823 Chr.20: 21162390-21162390 on Build GRCh38
rs374746995 Chr.20: 21162275-21162275 on Build GRCh38
rs983896453 Chr.20: 21162470-21162470 on Build GRCh38
rs183136126 Chr.20: 21162462-21162462 on Build GRCh38
rs750267893 Chr.20: 21162294-21162294 on Build GRCh38
rs200799486 Chr.20: 21162379-21162379 on Build GRCh38
rs756489931 Chr.20: 21162403-21162403 on Build GRCh38
rs760465667 Chr.20: 21162371-21162371 on Build GRCh38
rs116937124 Chr.20: 21162426-21162426 on Build GRCh38
rs368190969 Chr.20: 21162293-21162293 on Build GRCh38
rs768777591 Chr.20: 21162356-21162356 on Build GRCh38
rs972718508 Chr.20: 21162423-21162423 on Build GRCh38
rs772213549 Chr.20: 21162418-21162418 on Build GRCh38
rs778340669 Chr.20: 21162306-21162306 on Build GRCh38
rs770135261 Chr.20: 21162342-21162342 on Build GRCh38
rs772949553 Chr.20: 21162369-21162369 on Build GRCh38
rs752190640 Chr.20: 21162469-21162469 on Build GRCh38
rs769092674 Chr.20: 21162411-21162411 on Build GRCh38
rs766168725 Chr.20: 21162372-21162372 on Build GRCh38
rs369835898 Chr.20: 21162323-21162323 on Build GRCh38
rs747794715 Chr.20: 21162312-21162312 on Build GRCh38
rs141086448 Chr.20: 21162329-21162329 on Build GRCh38
rs35460260 Chr.20: 21162394-21162394 on Build GRCh38
rs753217273 Chr.20: 21162300-21162300 on Build GRCh38
rs781021625 Chr.20: 21162475-21162476 on Build GRCh38
rs201376938 Chr.20: 21162382-21162382 on Build GRCh38
rs754527224 Chr.20: 21162301-21162301 on Build GRCh38
rs748256445 Chr.20: 21162415-21162415 on Build GRCh38
rs746178337 Chr.20: 21162402-21162402 on Build GRCh38
rs185896476 Chr.20: 21162343-21162343 on Build GRCh38
rs190940090 Chr.20: 21162419-21162419 on Build GRCh38
rs781651685 Chr.20: 21162324-21162324 on Build GRCh38
rs754934073 Chr.20: 21162381-21162381 on Build GRCh38
rs780351048 Chr.20: 21162404-21162404 on Build GRCh38
rs369308735 Chr.20: 21162457-21162457 on Build GRCh38
rs961011642 Chr.20: 21162420-21162420 on Build GRCh38
rs377126281 Chr.20: 21162313-21162313 on Build GRCh38
rs762000568 Chr.20: 21162363-21162363 on Build GRCh38
rs758087503 Chr.20: 21162391-21162391 on Build GRCh38
rs372338345 Chr.20: 21162295-21162295 on Build GRCh38
rs774365618 Chr.20: 21162412-21162412 on Build GRCh38
rs1014842498 Chr.20: 21162358-21162358 on Build GRCh38
rs757151632 Chr.20: 21162337-21162338 on Build GRCh38
rs767747910 Chr.20: 21162367-21162367 on Build GRCh38
rs191064527 Chr.20: 21162357-21162357 on Build GRCh38
rs866489794 Chr.20: 21162398-21162398 on Build GRCh38
rs763676931 Chr.20: 21162318-21162318 on Build GRCh38
rs1048616045 Chr.20: 21162289-21162289 on Build GRCh38
rs185395297 Chr.20: 21162407-21162407 on Build GRCh38
rs776325775 Chr.20: 21162430-21162430 on Build GRCh38

View Full Product Details