Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137426265-137426526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOXA1
Entrez Gene ID
10811
Gene Name
NADPH oxidase activator 1
Gene Aliases
NY-CO-31, SDCCAG31, p51NOX
UniGene
Hs.495554
Gene Chromosome Location
Chr.9: 137423372-137434406 on Build GRCh38
Transcripts
NM_001256067.1 NM_001256068.1 NM_006647.1 XM_011518158.2 XM_011518159.1 XM_011518160.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5021660 c.260+10G>C p.? Chr.9: 137426340-137426340 on Build GRCh38
3367655 c.260+1G>A p.? Chr.9: 137426331-137426331 on Build GRCh38
4822088 c.202G>A p.D68N Chr.9: 137426272-137426272 on Build GRCh38
1107105 c.215C>T p.A72V Chr.9: 137426285-137426285 on Build GRCh38
6414823 c.226T>C p.F76L Chr.9: 137426296-137426296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962217517 Chr.9: 137426390-137426390 on Build GRCh38
rs970887392 Chr.9: 137426458-137426458 on Build GRCh38
rs72765154 Chr.9: 137426340-137426340 on Build GRCh38
rs760837435 Chr.9: 137426372-137426372 on Build GRCh38
rs114490343 Chr.9: 137426325-137426325 on Build GRCh38
rs927968051 Chr.9: 137426448-137426448 on Build GRCh38
rs538394742 Chr.9: 137426504-137426504 on Build GRCh38
rs929037968 Chr.9: 137426493-137426493 on Build GRCh38
rs763726979 Chr.9: 137426364-137426364 on Build GRCh38
rs920866532 Chr.9: 137426399-137426399 on Build GRCh38
rs904470298 Chr.9: 137426491-137426491 on Build GRCh38
rs766617824 Chr.9: 137426374-137426374 on Build GRCh38
rs28563072 Chr.9: 137426492-137426492 on Build GRCh38
rs959927008 Chr.9: 137426434-137426434 on Build GRCh38
rs1025606306 Chr.9: 137426452-137426452 on Build GRCh38
rs1052530537 Chr.9: 137426456-137426456 on Build GRCh38
rs548630963 Chr.9: 137426412-137426412 on Build GRCh38
rs758678542 Chr.9: 137426302-137426302 on Build GRCh38
rs376185485 Chr.9: 137426363-137426363 on Build GRCh38
rs945910509 Chr.9: 137426484-137426484 on Build GRCh38
rs756331723 Chr.9: 137426285-137426285 on Build GRCh38
rs753935714 Chr.9: 137426286-137426286 on Build GRCh38
rs746595589 Chr.9: 137426303-137426303 on Build GRCh38
rs891628123 Chr.9: 137426467-137426467 on Build GRCh38
rs369411282 Chr.9: 137426287-137426287 on Build GRCh38
rs774618380 Chr.9: 137426327-137426327 on Build GRCh38
rs149859430 Chr.9: 137426319-137426319 on Build GRCh38
rs562784135 Chr.9: 137426366-137426366 on Build GRCh38
rs770724744 Chr.9: 137426311-137426311 on Build GRCh38
rs146234834 Chr.9: 137426490-137426490 on Build GRCh38
rs566532329 Chr.9: 137426503-137426503 on Build GRCh38
rs59438560 Chr.9: 137426476-137426477 on Build GRCh38
rs1032343407 Chr.9: 137426420-137426420 on Build GRCh38
rs145312088 Chr.9: 137426475-137426476 on Build GRCh38
rs116517698 Chr.9: 137426443-137426443 on Build GRCh38
rs748738415 Chr.9: 137426298-137426298 on Build GRCh38
rs201743491 Chr.9: 137426465-137426475 on Build GRCh38
rs1000893816 Chr.9: 137426396-137426396 on Build GRCh38
rs532029184 Chr.9: 137426402-137426402 on Build GRCh38
rs565800104 Chr.9: 137426294-137426294 on Build GRCh38
rs745405690 Chr.9: 137426322-137426322 on Build GRCh38

View Full Product Details