Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45922944-45923175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARGE2
Entrez Gene ID
120071
Gene Name
LARGE xylosyl- and glucuronyltransferase 2
Gene Aliases
GYLTL1B, PP5656
UniGene
Hs.86543
Gene Chromosome Location
Chr.11: 45921621-45929096 on Build GRCh38
Transcripts
NM_001300721.1 NM_001300722.1 NM_152312.4 XM_006718140.1 XM_006718141.3 XM_011519886.1 XM_011519888.1 XM_011519889.1 XM_011519890.1 XM_011519891.1 XM_011519892.2 XM_011519893.1 XM_017017173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5446903 c.158T>C p.M53T Chr.11: 45923040-45923040 on Build GRCh38
5716508 c.169C>T p.P57S Chr.11: 45923051-45923051 on Build GRCh38
6365502 c.221C>T p.A74V Chr.11: 45923103-45923103 on Build GRCh38
6207737 c.285G>A p.E95E Chr.11: 45923167-45923167 on Build GRCh38
6865694 c.209G>A p.G70E Chr.11: 45923091-45923091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934687800 Chr.11: 45923086-45923086 on Build GRCh38
rs944815017 Chr.11: 45923131-45923131 on Build GRCh38
rs891485767 Chr.11: 45923055-45923055 on Build GRCh38
rs973955006 Chr.11: 45923142-45923143 on Build GRCh38
rs1029700080 Chr.11: 45923050-45923050 on Build GRCh38
rs769760213 Chr.11: 45922963-45922963 on Build GRCh38
rs776105032 Chr.11: 45923103-45923103 on Build GRCh38
rs900756151 Chr.11: 45922965-45922965 on Build GRCh38
rs932537493 Chr.11: 45923006-45923006 on Build GRCh38
rs1003946628 Chr.11: 45923000-45923000 on Build GRCh38
rs959857514 Chr.11: 45923029-45923029 on Build GRCh38
rs1022581226 Chr.11: 45923090-45923090 on Build GRCh38
rs970014306 Chr.11: 45923046-45923046 on Build GRCh38
rs768184108 Chr.11: 45923072-45923072 on Build GRCh38
rs543863960 Chr.11: 45923045-45923045 on Build GRCh38
rs1039298253 Chr.11: 45922964-45922964 on Build GRCh38
rs1013644763 Chr.11: 45923124-45923124 on Build GRCh38
rs17853729 Chr.11: 45922991-45922991 on Build GRCh38
rs773542378 Chr.11: 45922978-45922978 on Build GRCh38
rs904011062 Chr.11: 45923102-45923102 on Build GRCh38
rs550978364 Chr.11: 45923109-45923109 on Build GRCh38
rs1039225267 Chr.11: 45923136-45923136 on Build GRCh38
rs961078988 Chr.11: 45922977-45922977 on Build GRCh38
rs979148380 Chr.11: 45923047-45923047 on Build GRCh38
rs1049938531 Chr.11: 45923034-45923034 on Build GRCh38
rs992623801 Chr.11: 45923041-45923041 on Build GRCh38
rs1026662890 Chr.11: 45922990-45922990 on Build GRCh38
rs925959444 Chr.11: 45923078-45923078 on Build GRCh38
rs1015532733 Chr.11: 45923128-45923128 on Build GRCh38
rs996880632 Chr.11: 45922969-45922969 on Build GRCh38
rs777467056 Chr.11: 45923143-45923145 on Build GRCh38
rs973987559 Chr.11: 45922997-45922997 on Build GRCh38
rs771213705 Chr.11: 45922999-45922999 on Build GRCh38
rs912021514 Chr.11: 45923114-45923114 on Build GRCh38
rs199993922 Chr.11: 45923003-45923003 on Build GRCh38
rs1034271472 Chr.11: 45923106-45923106 on Build GRCh38

View Full Product Details