Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110875596-110875845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPAR1
Entrez Gene ID
1902
Gene Name
lysophosphatidic acid receptor 1
Gene Aliases
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
UniGene
Hs.126667
Gene Chromosome Location
Chr.9: 110873252-111039259 on Build GRCh38
Transcripts
NM_001401.3 NM_057159.2 XM_005251781.3 XM_005251782.3 XM_017014383.1 XM_017014384.1 XM_017014385.1 XM_017014386.1 XM_017014387.1 XM_017014388.1 XM_017014389.1 XM_017014390.1 XM_017014391.1 XM_017014392.1 XM_017014393.1 XM_017014394.1 XM_017014395.1 XM_017014396.1 XM_017014397.1 XM_017014398.1 XM_017014399.1 XM_017014400.1 XM_017014401.1 XM_017014402.1 XM_017014403.1 XM_017014404.1 XM_017014405.1 XM_017014406.1 XM_017014407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1104011 c.862G>A p.D288N Chr.9: 110875654-110875654 on Build GRCh38
3169728 c.843C>T p.D281D Chr.9: 110875673-110875673 on Build GRCh38
5559161 c.831G>A p.L277L Chr.9: 110875685-110875685 on Build GRCh38
7152837 c.911C>G p.S304C Chr.9: 110875605-110875605 on Build GRCh38
3925928 c.895C>T p.L299F Chr.9: 110875621-110875621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141775122 Chr.9: 110875655-110875655 on Build GRCh38
rs143615495 Chr.9: 110875648-110875648 on Build GRCh38
rs748360332 Chr.9: 110875715-110875715 on Build GRCh38
rs1023749673 Chr.9: 110875691-110875691 on Build GRCh38
rs760369860 Chr.9: 110875736-110875736 on Build GRCh38
rs149818114 Chr.9: 110875652-110875652 on Build GRCh38
rs757186566 Chr.9: 110875661-110875661 on Build GRCh38
rs1008968141 Chr.9: 110875796-110875796 on Build GRCh38
rs753785201 Chr.9: 110875659-110875659 on Build GRCh38
rs756216873 Chr.9: 110875718-110875718 on Build GRCh38
rs138203587 Chr.9: 110875697-110875697 on Build GRCh38
rs761314836 Chr.9: 110875702-110875703 on Build GRCh38
rs190849517 Chr.9: 110875788-110875788 on Build GRCh38
rs746236065 Chr.9: 110875728-110875728 on Build GRCh38
rs549709898 Chr.9: 110875787-110875787 on Build GRCh38
rs763552163 Chr.9: 110875738-110875738 on Build GRCh38
rs182493473 Chr.9: 110875790-110875790 on Build GRCh38
rs774955356 Chr.9: 110875726-110875726 on Build GRCh38
rs377562103 Chr.9: 110875729-110875729 on Build GRCh38
rs771244987 Chr.9: 110875643-110875643 on Build GRCh38
rs376237745 Chr.9: 110875710-110875710 on Build GRCh38
rs373021969 Chr.9: 110875671-110875671 on Build GRCh38
rs929261788 Chr.9: 110875775-110875775 on Build GRCh38
rs767305047 Chr.9: 110875651-110875651 on Build GRCh38
rs771616724 Chr.9: 110875646-110875646 on Build GRCh38
rs369322885 Chr.9: 110875669-110875669 on Build GRCh38
rs370157821 Chr.9: 110875732-110875732 on Build GRCh38
rs761188418 Chr.9: 110875745-110875745 on Build GRCh38
rs145823038 Chr.9: 110875654-110875654 on Build GRCh38
rs771487128 Chr.9: 110875724-110875724 on Build GRCh38
rs776182611 Chr.9: 110875741-110875741 on Build GRCh38
rs918520630 Chr.9: 110875748-110875748 on Build GRCh38
rs142862362 Chr.9: 110875719-110875719 on Build GRCh38
rs781239230 Chr.9: 110875752-110875753 on Build GRCh38
rs148990722 Chr.9: 110875673-110875673 on Build GRCh38
rs765164437 Chr.9: 110875764-110875764 on Build GRCh38

View Full Product Details