Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 33735135-33735408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPAS3
Entrez Gene ID
64067
Gene Name
neuronal PAS domain protein 3
Gene Aliases
MOP6, PASD6, bHLHe12
UniGene
Hs.657892
Gene Chromosome Location
Chr.14: 32934785-33804176 on Build GRCh38
Transcripts
NM_001164749.1 NM_001165893.1 NM_022123.2 NM_173159.2 XM_005267991.3 XM_005267992.3 XM_011537067.2 XM_011537069.2 XM_011537070.2 XM_011537071.2 XM_011537072.2 XM_017021582.1 XM_017021583.1 XM_017021584.1 XM_017021585.1 XM_017021586.1 XM_017021587.1 XM_017021588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5848044 c.743C>T p.S248L Chr.14: 33735319-33735319 on Build GRCh38
7191762 c.719C>A p.T240N Chr.14: 33735295-33735295 on Build GRCh38
5565073 c.781C>T p.R261C Chr.14: 33735261-33735261 on Build GRCh38
1707256 c.705G>A p.M235I Chr.14: 33735281-33735281 on Build GRCh38
5825249 c.686G>A p.R229H Chr.14: 33735262-33735262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758320179 Chr.14: 33735350-33735350 on Build GRCh38
rs368561940 Chr.14: 33735302-33735302 on Build GRCh38
rs777536559 Chr.14: 33735189-33735189 on Build GRCh38
rs777736607 Chr.14: 33735369-33735369 on Build GRCh38
rs769853726 Chr.14: 33735364-33735364 on Build GRCh38
rs755562964 Chr.14: 33735163-33735163 on Build GRCh38
rs763097646 Chr.14: 33735272-33735272 on Build GRCh38
rs973446301 Chr.14: 33735235-33735235 on Build GRCh38
rs778705581 Chr.14: 33735218-33735218 on Build GRCh38
rs780033019 Chr.14: 33735351-33735351 on Build GRCh38
rs752131671 Chr.14: 33735335-33735335 on Build GRCh38
rs745769093 Chr.14: 33735237-33735237 on Build GRCh38
rs191574245 Chr.14: 33735345-33735345 on Build GRCh38
rs750269970 Chr.14: 33735348-33735348 on Build GRCh38
rs778650810 Chr.14: 33735347-33735347 on Build GRCh38
rs748317339 Chr.14: 33735256-33735256 on Build GRCh38
rs142278370 Chr.14: 33735284-33735284 on Build GRCh38
rs368336896 Chr.14: 33735380-33735380 on Build GRCh38
rs950848692 Chr.14: 33735262-33735262 on Build GRCh38
rs773415582 Chr.14: 33735260-33735260 on Build GRCh38
rs757022778 Chr.14: 33735208-33735208 on Build GRCh38
rs774886036 Chr.14: 33735296-33735296 on Build GRCh38
rs763529607 Chr.14: 33735341-33735341 on Build GRCh38
rs748900840 Chr.14: 33735203-33735203 on Build GRCh38
rs749483985 Chr.14: 33735370-33735370 on Build GRCh38
rs148708760 Chr.14: 33735164-33735164 on Build GRCh38
rs756938009 Chr.14: 33735346-33735346 on Build GRCh38
rs963381788 Chr.14: 33735229-33735229 on Build GRCh38
rs571164219 Chr.14: 33735194-33735195 on Build GRCh38
rs1013124034 Chr.14: 33735201-33735201 on Build GRCh38
rs892045179 Chr.14: 33735194-33735194 on Build GRCh38
rs769905118 Chr.14: 33735257-33735257 on Build GRCh38
rs151297906 Chr.14: 33735252-33735252 on Build GRCh38
rs961274871 Chr.14: 33735359-33735359 on Build GRCh38
rs375251895 Chr.14: 33735253-33735253 on Build GRCh38
rs201700880 Chr.14: 33735184-33735184 on Build GRCh38
rs140556633 Chr.14: 33735322-33735322 on Build GRCh38
rs947057537 Chr.14: 33735366-33735366 on Build GRCh38
rs112057025 Chr.14: 33735340-33735340 on Build GRCh38
rs752289450 Chr.14: 33735178-33735178 on Build GRCh38
rs1024102946 Chr.14: 33735376-33735376 on Build GRCh38
rs73258990 Chr.14: 33735356-33735356 on Build GRCh38
rs149680595 Chr.14: 33735329-33735329 on Build GRCh38
rs772039656 Chr.14: 33735240-33735240 on Build GRCh38

View Full Product Details