Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3360731-3360956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF195
Entrez Gene ID
7748
Gene Name
zinc finger protein 195
Gene Aliases
HRF1, ZNFP104
UniGene
Hs.386294
Gene Chromosome Location
Chr.11: 3357927-3379222 on Build GRCh38
Transcripts
NM_001130519.2 NM_001130520.2 NM_001242841.1 NM_001242842.1 NM_001242843.1 NM_001256823.1 NM_001256824.1 NM_001256825.1 NM_007152.4 NR_040083.1 NR_046381.1 NR_046382.1 XM_006718306.1 XM_011520350.2 XM_011520351.2 XM_011520352.2 XM_011520354.2 XM_017018260.1 XM_017018261.1 XM_017018262.1 XM_017018263.1 XM_017018264.1 XM_017018265.1 XM_017018266.1 XM_017018267.1

SNP Detail

SNP ID Chromosome Location
rs374162006 Chr.11: 3360780-3360780 on Build GRCh38
rs770137999 Chr.11: 3360830-3360830 on Build GRCh38
rs780265114 Chr.11: 3360844-3360844 on Build GRCh38
rs913652973 Chr.11: 3360793-3360793 on Build GRCh38
rs373123026 Chr.11: 3360766-3360766 on Build GRCh38
rs574472009 Chr.11: 3360852-3360852 on Build GRCh38
rs139352237 Chr.11: 3360832-3360832 on Build GRCh38
rs569579741 Chr.11: 3360829-3360829 on Build GRCh38
rs552985852 Chr.11: 3360851-3360851 on Build GRCh38
rs769758077 Chr.11: 3360894-3360894 on Build GRCh38
rs144090930 Chr.11: 3360836-3360836 on Build GRCh38
rs575989248 Chr.11: 3360886-3360886 on Build GRCh38
rs900571870 Chr.11: 3360786-3360786 on Build GRCh38
rs1024987800 Chr.11: 3360915-3360915 on Build GRCh38
rs981116585 Chr.11: 3360867-3360867 on Build GRCh38
rs748467521 Chr.11: 3360768-3360768 on Build GRCh38
rs945070239 Chr.11: 3360800-3360800 on Build GRCh38
rs184239505 Chr.11: 3360840-3360840 on Build GRCh38
rs550583352 Chr.11: 3360824-3360824 on Build GRCh38
rs6578335 Chr.11: 3360839-3360839 on Build GRCh38
rs113722759 Chr.11: 3360858-3360858 on Build GRCh38
rs557032329 Chr.11: 3360869-3360869 on Build GRCh38
rs1051426833 Chr.11: 3360810-3360810 on Build GRCh38
rs947579798 Chr.11: 3360779-3360779 on Build GRCh38

View Full Product Details