Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55025008-55025235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTN4
Entrez Gene ID
57142
Gene Name
reticulon 4
Gene Aliases
ASY, NI220/250, NOGO, NOGO-A, NOGOC, NSP, NSP-CL, Nbla00271, Nbla10545, Nogo-B, Nogo-C, RTN-X, RTN4-A, RTN4-B1, RTN4-B2, RTN4-C
UniGene
Hs.637850
Gene Chromosome Location
Chr.2: 54972187-55137831 on Build GRCh38
Transcripts
NM_001321859.1 NM_001321860.1 NM_001321861.1 NM_001321862.1 NM_001321863.1 NM_001321904.1 NM_020532.4 NM_153828.2 NM_207520.1 NM_207521.1 NR_135829.1 NR_135830.1 XM_005264434.3 XM_017004518.1 XM_017004519.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4540718 c.2889G>A p.E963E Chr.2: 55025210-55025210 on Build GRCh38
6421079 c.2977T>C p.S993P Chr.2: 55025122-55025122 on Build GRCh38
4747278 c.2274delA p.I758fs*4 Chr.2: 55025207-55025207 on Build GRCh38
4827164 c.2944C>T p.P982S Chr.2: 55025155-55025155 on Build GRCh38
6235916 c.2332G>A p.D778N Chr.2: 55025149-55025149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78508174 Chr.2: 55025041-55025041 on Build GRCh38
rs368860803 Chr.2: 55025174-55025174 on Build GRCh38
rs778437228 Chr.2: 55025053-55025053 on Build GRCh38
rs546099669 Chr.2: 55025115-55025115 on Build GRCh38
rs201732018 Chr.2: 55025147-55025147 on Build GRCh38
rs902313941 Chr.2: 55025112-55025112 on Build GRCh38
rs768788957 Chr.2: 55025187-55025187 on Build GRCh38
rs200210000 Chr.2: 55025098-55025098 on Build GRCh38
rs764839209 Chr.2: 55025140-55025140 on Build GRCh38
rs200503618 Chr.2: 55025209-55025209 on Build GRCh38
rs201070007 Chr.2: 55025071-55025071 on Build GRCh38
rs111958278 Chr.2: 55025123-55025123 on Build GRCh38
rs764933644 Chr.2: 55025208-55025208 on Build GRCh38
rs906823854 Chr.2: 55025188-55025188 on Build GRCh38
rs764683289 Chr.2: 55025121-55025121 on Build GRCh38
rs777686956 Chr.2: 55025145-55025145 on Build GRCh38
rs757205740 Chr.2: 55025148-55025148 on Build GRCh38
rs745687944 Chr.2: 55025150-55025150 on Build GRCh38
rs559625826 Chr.2: 55025077-55025077 on Build GRCh38
rs201229603 Chr.2: 55025116-55025116 on Build GRCh38
rs144381198 Chr.2: 55025048-55025048 on Build GRCh38
rs767865946 Chr.2: 55025129-55025129 on Build GRCh38
rs759190599 Chr.2: 55025132-55025132 on Build GRCh38
rs749372472 Chr.2: 55025182-55025182 on Build GRCh38
rs757374131 Chr.2: 55025063-55025063 on Build GRCh38
rs776298868 Chr.2: 55025091-55025091 on Build GRCh38
rs767412022 Chr.2: 55025094-55025094 on Build GRCh38
rs139414628 Chr.2: 55025125-55025125 on Build GRCh38
rs774585686 Chr.2: 55025190-55025190 on Build GRCh38
rs774430562 Chr.2: 55025060-55025060 on Build GRCh38
rs750775178 Chr.2: 55025131-55025131 on Build GRCh38
rs145032505 Chr.2: 55025149-55025149 on Build GRCh38
rs199769551 Chr.2: 55025117-55025117 on Build GRCh38
rs200950640 Chr.2: 55025185-55025185 on Build GRCh38
rs546003683 Chr.2: 55025076-55025076 on Build GRCh38
rs199592685 Chr.2: 55025198-55025198 on Build GRCh38
rs13401937 Chr.2: 55025078-55025078 on Build GRCh38
rs752232361 Chr.2: 55025143-55025143 on Build GRCh38
rs200491416 Chr.2: 55025070-55025070 on Build GRCh38
rs199774136 Chr.2: 55025158-55025158 on Build GRCh38
rs372405619 Chr.2: 55025104-55025104 on Build GRCh38
rs762361993 Chr.2: 55025191-55025191 on Build GRCh38
rs972539939 Chr.2: 55025166-55025166 on Build GRCh38
rs199645396 Chr.2: 55025214-55025214 on Build GRCh38
rs760996053 Chr.2: 55025202-55025202 on Build GRCh38
rs773597274 Chr.2: 55025199-55025199 on Build GRCh38
rs201421748 Chr.2: 55025192-55025192 on Build GRCh38
rs763172993 Chr.2: 55025119-55025119 on Build GRCh38
rs185241265 Chr.2: 55025058-55025058 on Build GRCh38
rs201924443 Chr.2: 55025213-55025213 on Build GRCh38
rs757964147 Chr.2: 55025144-55025144 on Build GRCh38
rs747645565 Chr.2: 55025054-55025054 on Build GRCh38

View Full Product Details