Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 39716877-39717104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM178A
Entrez Gene ID
130733
Gene Name
transmembrane protein 178A
Gene Aliases
TMEM178
UniGene
Hs.40808
Gene Chromosome Location
Chr.2: 39665087-39717963 on Build GRCh38
Transcripts
NM_001167959.1 NM_152390.2 XM_005264144.1 XM_005264145.1 XM_017003369.1 XM_017003370.1 XM_017003371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3581546 c.671C>T p.S224F Chr.2: 39717028-39717028 on Build GRCh38
7411095 c.686C>T p.A229V Chr.2: 39717043-39717043 on Build GRCh38
721003 c.653G>A p.G218E Chr.2: 39717010-39717010 on Build GRCh38
5397010 c.689C>A p.A230D Chr.2: 39717046-39717046 on Build GRCh38
1020906 c.699G>A p.S233S Chr.2: 39717056-39717056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747145533 Chr.2: 39717044-39717044 on Build GRCh38
rs942529412 Chr.2: 39716978-39716978 on Build GRCh38
rs889487585 Chr.2: 39716924-39716924 on Build GRCh38
rs780403604 Chr.2: 39717042-39717042 on Build GRCh38
rs762535118 Chr.2: 39716969-39716969 on Build GRCh38
rs778307347 Chr.2: 39717000-39717000 on Build GRCh38
rs769722792 Chr.2: 39717059-39717059 on Build GRCh38
rs764947451 Chr.2: 39716962-39716962 on Build GRCh38
rs759110252 Chr.2: 39717075-39717075 on Build GRCh38
rs370900847 Chr.2: 39717032-39717032 on Build GRCh38
rs773977856 Chr.2: 39717074-39717074 on Build GRCh38
rs201281373 Chr.2: 39717012-39717012 on Build GRCh38
rs371628776 Chr.2: 39716913-39716913 on Build GRCh38
rs748048353 Chr.2: 39717056-39717056 on Build GRCh38
rs766013549 Chr.2: 39716975-39716975 on Build GRCh38
rs776786440 Chr.2: 39717055-39717055 on Build GRCh38
rs115432122 Chr.2: 39716927-39716927 on Build GRCh38
rs754459713 Chr.2: 39716993-39716993 on Build GRCh38
rs750173301 Chr.2: 39716965-39716965 on Build GRCh38
rs754335043 Chr.2: 39717015-39717015 on Build GRCh38
rs758590980 Chr.2: 39717041-39717041 on Build GRCh38
rs751107888 Chr.2: 39716992-39716992 on Build GRCh38
rs199591656 Chr.2: 39717043-39717043 on Build GRCh38
rs372186330 Chr.2: 39717071-39717071 on Build GRCh38
rs2540229 Chr.2: 39716915-39716915 on Build GRCh38
rs772909667 Chr.2: 39717068-39717068 on Build GRCh38
rs774066397 Chr.2: 39717078-39717078 on Build GRCh38
rs768433672 Chr.2: 39717011-39717011 on Build GRCh38
rs1019396189 Chr.2: 39716948-39716948 on Build GRCh38
rs141928270 Chr.2: 39717045-39717045 on Build GRCh38
rs200061855 Chr.2: 39717026-39717026 on Build GRCh38
rs889169067 Chr.2: 39716949-39716949 on Build GRCh38

View Full Product Details