Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 40265604-40265833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNRC6B
Entrez Gene ID
23112
Gene Name
trinucleotide repeat containing 6B
Gene Aliases
-
UniGene
Hs.372082
Gene Chromosome Location
Chr.22: 40044817-40335808 on Build GRCh38
Transcripts
NM_001024843.1 NM_001162501.1 NM_015088.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5177503 c.1461C>T p.G487G Chr.22: 40265691-40265691 on Build GRCh38
5990531 c.1411G>T p.D471Y Chr.22: 40265641-40265641 on Build GRCh38
4753472 c.1508A>G p.Q503R Chr.22: 40265696-40265696 on Build GRCh38
1593065 c.1476G>A p.R492R Chr.22: 40265664-40265664 on Build GRCh38
3740435 c.1623T>G p.G541G Chr.22: 40265811-40265811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756271910 Chr.22: 40265722-40265722 on Build GRCh38
rs780999754 Chr.22: 40265720-40265720 on Build GRCh38
rs199666343 Chr.22: 40265668-40265668 on Build GRCh38
rs908293511 Chr.22: 40265639-40265639 on Build GRCh38
rs989350441 Chr.22: 40265761-40265761 on Build GRCh38
rs193065497 Chr.22: 40265734-40265734 on Build GRCh38
rs768312489 Chr.22: 40265669-40265669 on Build GRCh38
rs769670762 Chr.22: 40265696-40265696 on Build GRCh38
rs376115865 Chr.22: 40265689-40265689 on Build GRCh38
rs374041253 Chr.22: 40265712-40265712 on Build GRCh38
rs752924057 Chr.22: 40265719-40265719 on Build GRCh38
rs756448992 Chr.22: 40265646-40265646 on Build GRCh38
rs762586017 Chr.22: 40265810-40265810 on Build GRCh38
rs370999187 Chr.22: 40265745-40265745 on Build GRCh38
rs767871405 Chr.22: 40265716-40265716 on Build GRCh38
rs17001767 Chr.22: 40265780-40265780 on Build GRCh38
rs779609225 Chr.22: 40265664-40265664 on Build GRCh38
rs757097943 Chr.22: 40265688-40265688 on Build GRCh38
rs746545703 Chr.22: 40265759-40265759 on Build GRCh38
rs772860753 Chr.22: 40265699-40265699 on Build GRCh38
rs898542626 Chr.22: 40265635-40265635 on Build GRCh38
rs943783467 Chr.22: 40265648-40265648 on Build GRCh38
rs753122544 Chr.22: 40265645-40265645 on Build GRCh38
rs370276937 Chr.22: 40265705-40265705 on Build GRCh38
rs776218575 Chr.22: 40265670-40265670 on Build GRCh38
rs766220961 Chr.22: 40265708-40265708 on Build GRCh38
rs371005172 Chr.22: 40265657-40265657 on Build GRCh38
rs375620018 Chr.22: 40265779-40265779 on Build GRCh38
rs373211790 Chr.22: 40265661-40265661 on Build GRCh38
rs749860211 Chr.22: 40265658-40265658 on Build GRCh38
rs146306455 Chr.22: 40265725-40265725 on Build GRCh38
rs768116213 Chr.22: 40265764-40265764 on Build GRCh38
rs780650568 Chr.22: 40265771-40265771 on Build GRCh38
rs773124703 Chr.22: 40265795-40265795 on Build GRCh38
rs751692063 Chr.22: 40265711-40265711 on Build GRCh38
rs769317646 Chr.22: 40265793-40265793 on Build GRCh38

View Full Product Details