Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 98217827-98218070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TECPR1
Entrez Gene ID
25851
Gene Name
tectonin beta-propeller repeat containing 1
Gene Aliases
-
UniGene
Hs.592281
Gene Chromosome Location
Chr.7: 98215443-98252251 on Build GRCh38
Transcripts
NM_015395.2 XM_005250253.3 XM_017011937.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4996877 c.3238T>C p.C1080R Chr.7: 98217965-98217965 on Build GRCh38
6189315 c.3176G>A p.R1059H Chr.7: 98218027-98218027 on Build GRCh38
1093482 c.3163A>G p.N1055D Chr.7: 98218040-98218040 on Build GRCh38
4779074 c.3211A>G p.S1071G Chr.7: 98217992-98217992 on Build GRCh38
5835483 c.3200delC p.P1067fs*26 Chr.7: 98218003-98218003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750776780 Chr.7: 98217980-98217980 on Build GRCh38
rs780778151 Chr.7: 98218004-98218004 on Build GRCh38
rs377595090 Chr.7: 98218002-98218002 on Build GRCh38
rs373155446 Chr.7: 98217981-98217981 on Build GRCh38
rs762851894 Chr.7: 98217931-98217931 on Build GRCh38
rs909317052 Chr.7: 98217924-98217924 on Build GRCh38
rs769713156 Chr.7: 98217914-98217914 on Build GRCh38
rs918167021 Chr.7: 98217859-98217859 on Build GRCh38
rs773051091 Chr.7: 98217926-98217926 on Build GRCh38
rs546176846 Chr.7: 98217962-98217962 on Build GRCh38
rs368893258 Chr.7: 98217851-98217851 on Build GRCh38
rs762019037 Chr.7: 98217968-98217968 on Build GRCh38
rs1048162541 Chr.7: 98217908-98217908 on Build GRCh38
rs760768292 Chr.7: 98217959-98217959 on Build GRCh38
rs1048284082 Chr.7: 98218046-98218046 on Build GRCh38
rs770873755 Chr.7: 98217945-98217945 on Build GRCh38
rs754547055 Chr.7: 98218003-98218003 on Build GRCh38
rs138522081 Chr.7: 98218015-98218015 on Build GRCh38
rs766939393 Chr.7: 98217988-98217988 on Build GRCh38
rs374604046 Chr.7: 98218014-98218014 on Build GRCh38
rs747872474 Chr.7: 98218005-98218005 on Build GRCh38
rs528019491 Chr.7: 98217961-98217961 on Build GRCh38
rs752208106 Chr.7: 98217858-98217858 on Build GRCh38
rs929332633 Chr.7: 98217873-98217873 on Build GRCh38
rs557294111 Chr.7: 98217911-98217920 on Build GRCh38
rs765578656 Chr.7: 98217969-98217969 on Build GRCh38
rs1001659499 Chr.7: 98218027-98218027 on Build GRCh38
rs1000879074 Chr.7: 98217865-98217865 on Build GRCh38
rs968501470 Chr.7: 98218024-98218024 on Build GRCh38
rs777825235 Chr.7: 98217990-98217990 on Build GRCh38
rs990930696 Chr.7: 98217920-98217920 on Build GRCh38
rs1032318396 Chr.7: 98217881-98217881 on Build GRCh38
rs774118240 Chr.7: 98217954-98217954 on Build GRCh38

View Full Product Details