Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46690783-46691056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDRD6
Entrez Gene ID
221400
Gene Name
tudor domain containing 6
Gene Aliases
CT41.2, NY-CO-45, SPATA36, TDR2, bA446F17.4
UniGene
Hs.656983
Gene Chromosome Location
Chr.6: 46687875-46704319 on Build GRCh38
Transcripts
NM_001010870.2 NM_001168359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7245148 c.2744A>C p.Q915P Chr.6: 46690872-46690872 on Build GRCh38
484155 c.2746A>C p.K916Q Chr.6: 46690874-46690874 on Build GRCh38
5996563 c.2765G>A p.C922Y Chr.6: 46690893-46690893 on Build GRCh38
1660199 c.2722G>T p.E908* Chr.6: 46690850-46690850 on Build GRCh38
76701 c.2926T>C p.F976L Chr.6: 46691054-46691054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753132766 Chr.6: 46690863-46690863 on Build GRCh38
rs61742452 Chr.6: 46690823-46690823 on Build GRCh38
rs915567099 Chr.6: 46690840-46690840 on Build GRCh38
rs888959717 Chr.6: 46691004-46691004 on Build GRCh38
rs756534523 Chr.6: 46690936-46690936 on Build GRCh38
rs768383972 Chr.6: 46690812-46690812 on Build GRCh38
rs766534661 Chr.6: 46690839-46690839 on Build GRCh38
rs751671231 Chr.6: 46690848-46690848 on Build GRCh38
rs762957682 Chr.6: 46690916-46690916 on Build GRCh38
rs553633867 Chr.6: 46691028-46691028 on Build GRCh38
rs1036343465 Chr.6: 46690907-46690907 on Build GRCh38
rs749134745 Chr.6: 46691012-46691012 on Build GRCh38
rs767818120 Chr.6: 46690930-46690930 on Build GRCh38
rs752733915 Chr.6: 46690908-46690908 on Build GRCh38
rs199823660 Chr.6: 46691019-46691019 on Build GRCh38
rs773023800 Chr.6: 46690900-46690900 on Build GRCh38
rs764586006 Chr.6: 46690937-46690937 on Build GRCh38
rs762915207 Chr.6: 46690836-46690836 on Build GRCh38
rs780797714 Chr.6: 46690969-46690969 on Build GRCh38
rs770934988 Chr.6: 46691015-46691015 on Build GRCh38
rs757794338 Chr.6: 46690948-46690948 on Build GRCh38
rs537058140 Chr.6: 46690867-46690867 on Build GRCh38
rs753018392 Chr.6: 46690935-46690935 on Build GRCh38
rs755253297 Chr.6: 46690856-46690856 on Build GRCh38
rs773253601 Chr.6: 46690831-46690831 on Build GRCh38
rs754519785 Chr.6: 46690968-46690968 on Build GRCh38
rs759794628 Chr.6: 46690929-46690929 on Build GRCh38
rs747918726 Chr.6: 46690894-46690894 on Build GRCh38
rs765467808 Chr.6: 46690871-46690872 on Build GRCh38
rs757834337 Chr.6: 46690868-46690868 on Build GRCh38
rs920155078 Chr.6: 46690963-46690963 on Build GRCh38
rs150117036 Chr.6: 46691020-46691020 on Build GRCh38
rs746551549 Chr.6: 46690954-46690954 on Build GRCh38
rs754231804 Chr.6: 46690943-46690943 on Build GRCh38
rs766298927 Chr.6: 46690919-46690919 on Build GRCh38
rs779381214 Chr.6: 46690949-46690949 on Build GRCh38
rs759699021 Chr.6: 46690814-46690817 on Build GRCh38
rs763133186 Chr.6: 46691023-46691023 on Build GRCh38
rs778111245 Chr.6: 46690866-46690866 on Build GRCh38
rs756593966 Chr.6: 46690864-46690864 on Build GRCh38
rs779630648 Chr.6: 46690879-46690879 on Build GRCh38
rs776364304 Chr.6: 46690893-46690893 on Build GRCh38
rs776417819 Chr.6: 46690817-46690817 on Build GRCh38
rs774197702 Chr.6: 46691016-46691016 on Build GRCh38
rs746576872 Chr.6: 46690880-46690880 on Build GRCh38
rs142686920 Chr.6: 46690985-46690985 on Build GRCh38
rs774517326 Chr.6: 46690921-46690921 on Build GRCh38
rs759486268 Chr.6: 46691030-46691030 on Build GRCh38
rs777640964 Chr.6: 46691009-46691009 on Build GRCh38
rs769620382 Chr.6: 46690898-46690898 on Build GRCh38
rs948179453 Chr.6: 46690862-46690862 on Build GRCh38
rs141480475 Chr.6: 46690882-46690882 on Build GRCh38
rs775851270 Chr.6: 46691021-46691021 on Build GRCh38
rs767951236 Chr.6: 46690861-46690861 on Build GRCh38
rs769413670 Chr.6: 46690991-46690991 on Build GRCh38

View Full Product Details