Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36699708-36699933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIM2
Entrez Gene ID
6493
Gene Name
single-minded family bHLH transcription factor 2
Gene Aliases
HMC13F06, HMC29C01, SIM, bHLHe15
UniGene
Hs.146186
Gene Chromosome Location
Chr.21: 36699117-36750219 on Build GRCh38
Transcripts
NM_005069.4 NM_009586.3 XM_017028442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030543 c.159C>T p.R53R Chr.21: 36699905-36699905 on Build GRCh38
5151115 c.170C>T p.P57L Chr.21: 36699916-36699916 on Build GRCh38
4185628 c.130C>G p.R44G Chr.21: 36699876-36699876 on Build GRCh38
5392880 c.169C>T p.P57S Chr.21: 36699915-36699915 on Build GRCh38
2844684 c.54C>A p.G18G Chr.21: 36699800-36699800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750361630 Chr.21: 36699906-36699906 on Build GRCh38
rs375415183 Chr.21: 36699821-36699821 on Build GRCh38
rs764264617 Chr.21: 36699881-36699881 on Build GRCh38
rs774836202 Chr.21: 36699734-36699734 on Build GRCh38
rs200229277 Chr.21: 36699800-36699800 on Build GRCh38
rs376678295 Chr.21: 36699799-36699799 on Build GRCh38
rs761019093 Chr.21: 36699878-36699878 on Build GRCh38
rs566219140 Chr.21: 36699752-36699752 on Build GRCh38
rs772347076 Chr.21: 36699865-36699865 on Build GRCh38
rs145240420 Chr.21: 36699791-36699791 on Build GRCh38
rs767710323 Chr.21: 36699742-36699742 on Build GRCh38
rs142269949 Chr.21: 36699899-36699899 on Build GRCh38
rs759831335 Chr.21: 36699740-36699740 on Build GRCh38
rs996124438 Chr.21: 36699876-36699876 on Build GRCh38
rs148750764 Chr.21: 36699836-36699836 on Build GRCh38
rs202090892 Chr.21: 36699827-36699827 on Build GRCh38
rs750179033 Chr.21: 36699809-36699809 on Build GRCh38
rs749781705 Chr.21: 36699839-36699839 on Build GRCh38
rs765158971 Chr.21: 36699903-36699903 on Build GRCh38
rs776913500 Chr.21: 36699887-36699887 on Build GRCh38
rs746201650 Chr.21: 36699861-36699861 on Build GRCh38
rs377481674 Chr.21: 36699866-36699866 on Build GRCh38
rs979374259 Chr.21: 36699732-36699732 on Build GRCh38
rs780742769 Chr.21: 36699838-36699838 on Build GRCh38
rs758135000 Chr.21: 36699810-36699810 on Build GRCh38
rs753796262 Chr.21: 36699768-36699768 on Build GRCh38
rs764319761 Chr.21: 36699760-36699760 on Build GRCh38
rs940139819 Chr.21: 36699847-36699847 on Build GRCh38
rs760789833 Chr.21: 36699756-36699756 on Build GRCh38
rs535903610 Chr.21: 36699859-36699859 on Build GRCh38
rs771411081 Chr.21: 36699845-36699845 on Build GRCh38

View Full Product Details