Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4817679-4817912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TICAM1
Entrez Gene ID
148022
Gene Name
toll like receptor adaptor molecule 1
Gene Aliases
IIAE6, MyD88-3, PRVTIRB, TICAM-1, TRIF
UniGene
Hs.29344
Gene Chromosome Location
Chr.19: 4815924-4831742 on Build GRCh38
Transcripts
NM_182919.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4188253 c.653T>C p.L218P Chr.19: 4817725-4817725 on Build GRCh38
6870748 c.583C>G p.R195G Chr.19: 4817795-4817795 on Build GRCh38
5015570 c.533T>C p.L178P Chr.19: 4817845-4817845 on Build GRCh38
5015571 c.530G>A p.S177N Chr.19: 4817848-4817848 on Build GRCh38
5056794 c.539G>A p.R180H Chr.19: 4817839-4817839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762194513 Chr.19: 4817825-4817825 on Build GRCh38
rs766336167 Chr.19: 4817752-4817752 on Build GRCh38
rs751395479 Chr.19: 4817760-4817760 on Build GRCh38
rs754152546 Chr.19: 4817730-4817730 on Build GRCh38
rs1044560697 Chr.19: 4817769-4817769 on Build GRCh38
rs762106344 Chr.19: 4817736-4817736 on Build GRCh38
rs866478664 Chr.19: 4817807-4817807 on Build GRCh38
rs562147335 Chr.19: 4817781-4817781 on Build GRCh38
rs375230364 Chr.19: 4817834-4817834 on Build GRCh38
rs759330259 Chr.19: 4817829-4817829 on Build GRCh38
rs369799199 Chr.19: 4817871-4817871 on Build GRCh38
rs772478656 Chr.19: 4817800-4817800 on Build GRCh38
rs760097887 Chr.19: 4817698-4817698 on Build GRCh38
rs768828649 Chr.19: 4817820-4817820 on Build GRCh38
rs772559700 Chr.19: 4817710-4817710 on Build GRCh38
rs745809710 Chr.19: 4817835-4817835 on Build GRCh38
rs940970222 Chr.19: 4817839-4817839 on Build GRCh38
rs373468408 Chr.19: 4817872-4817872 on Build GRCh38
rs756897138 Chr.19: 4817789-4817789 on Build GRCh38
rs763658275 Chr.19: 4817854-4817854 on Build GRCh38
rs755612385 Chr.19: 4817775-4817775 on Build GRCh38
rs141394423 Chr.19: 4817840-4817840 on Build GRCh38
rs753273529 Chr.19: 4817864-4817864 on Build GRCh38
rs780257437 Chr.19: 4817894-4817894 on Build GRCh38
rs747441149 Chr.19: 4817812-4817812 on Build GRCh38
rs895523368 Chr.19: 4817708-4817708 on Build GRCh38
rs760759367 Chr.19: 4817718-4817718 on Build GRCh38
rs775445910 Chr.19: 4817713-4817713 on Build GRCh38
rs765489270 Chr.19: 4817828-4817828 on Build GRCh38
rs756911879 Chr.19: 4817866-4817866 on Build GRCh38
rs776024318 Chr.19: 4817805-4817805 on Build GRCh38
rs752174873 Chr.19: 4817772-4817772 on Build GRCh38
rs758835511 Chr.19: 4817883-4817883 on Build GRCh38
rs767434262 Chr.19: 4817766-4817766 on Build GRCh38
rs754876060 Chr.19: 4817764-4817764 on Build GRCh38
rs746169578 Chr.19: 4817795-4817795 on Build GRCh38
rs146550489 Chr.19: 4817821-4817821 on Build GRCh38
rs200795596 Chr.19: 4817852-4817852 on Build GRCh38
rs779250040 Chr.19: 4817794-4817794 on Build GRCh38
rs764252931 Chr.19: 4817721-4817721 on Build GRCh38

View Full Product Details