Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 169743173-169743416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCTE3
Entrez Gene ID
6991
Gene Name
t-complex-associated-testis-expressed 3
Gene Aliases
TCTEX1D3
UniGene
Hs.733746
Gene Chromosome Location
Chr.6: 169725488-169751581 on Build GRCh38
Transcripts
NM_174910.1 XM_006715554.3 XM_011536093.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1076168 c.356A>G p.D119G Chr.6: 169743210-169743210 on Build GRCh38
6392413 c.350A>G p.Y117C Chr.6: 169743216-169743216 on Build GRCh38
4805340 c.365T>C p.F122S Chr.6: 169743201-169743201 on Build GRCh38
6393289 c.334C>A p.L112I Chr.6: 169743232-169743232 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1030024271 Chr.6: 169743323-169743324 on Build GRCh38
rs374351298 Chr.6: 169743325-169743325 on Build GRCh38
rs753294011 Chr.6: 169743270-169743270 on Build GRCh38
rs141929995 Chr.6: 169743207-169743207 on Build GRCh38
rs756305578 Chr.6: 169743261-169743261 on Build GRCh38
rs754512517 Chr.6: 169743358-169743358 on Build GRCh38
rs886570521 Chr.6: 169743215-169743215 on Build GRCh38
rs1029281509 Chr.6: 169743216-169743216 on Build GRCh38
rs755170983 Chr.6: 169743281-169743281 on Build GRCh38
rs115517841 Chr.6: 169743313-169743313 on Build GRCh38
rs13194101 Chr.6: 169743204-169743204 on Build GRCh38
rs780314093 Chr.6: 169743266-169743266 on Build GRCh38
rs750946935 Chr.6: 169743228-169743228 on Build GRCh38
rs948023725 Chr.6: 169743362-169743362 on Build GRCh38
rs751880193 Chr.6: 169743211-169743211 on Build GRCh38
rs779125750 Chr.6: 169743290-169743290 on Build GRCh38
rs920598639 Chr.6: 169743357-169743357 on Build GRCh38
rs753907605 Chr.6: 169743280-169743280 on Build GRCh38
rs763749953 Chr.6: 169743208-169743208 on Build GRCh38
rs762166347 Chr.6: 169743225-169743225 on Build GRCh38
rs973872731 Chr.6: 169743335-169743335 on Build GRCh38
rs370382795 Chr.6: 169743316-169743316 on Build GRCh38
rs980732582 Chr.6: 169743377-169743377 on Build GRCh38
rs954938215 Chr.6: 169743219-169743219 on Build GRCh38
rs927253851 Chr.6: 169743379-169743379 on Build GRCh38

View Full Product Details