Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 92094802-92095065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLCO3A1
Entrez Gene ID
28232
Gene Name
solute carrier organic anion transporter family member 3A1
Gene Aliases
OATP-D, OATP-RP3, OATP3A1, OATPD, OATPRP3, SLC21A11
UniGene
Hs.311187
Gene Chromosome Location
Chr.15: 91853708-92172435 on Build GRCh38
Transcripts
NM_001145044.1 NM_013272.3 NR_135775.1 XM_005254889.1 XM_005254891.2 XM_011521456.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
966661 c.684C>T p.C228C Chr.15: 92094918-92094918 on Build GRCh38
1478477 c.648A>T p.G216G Chr.15: 92094882-92094882 on Build GRCh38
4727956 c.728C>T p.A243V Chr.15: 92094962-92094962 on Build GRCh38
3505497 c.660G>A p.T220T Chr.15: 92094894-92094894 on Build GRCh38
5651045 c.715A>T p.I239F Chr.15: 92094949-92094949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754631677 Chr.15: 92094954-92094954 on Build GRCh38
rs12594058 Chr.15: 92094894-92094894 on Build GRCh38
rs751930192 Chr.15: 92094884-92094884 on Build GRCh38
rs77903937 Chr.15: 92094869-92094869 on Build GRCh38
rs764263237 Chr.15: 92094883-92094883 on Build GRCh38
rs750914605 Chr.15: 92095019-92095019 on Build GRCh38
rs147925181 Chr.15: 92094888-92094888 on Build GRCh38
rs747929610 Chr.15: 92094962-92094962 on Build GRCh38
rs377427714 Chr.15: 92095013-92095013 on Build GRCh38
rs114505280 Chr.15: 92095017-92095017 on Build GRCh38
rs762204398 Chr.15: 92095016-92095016 on Build GRCh38
rs887649066 Chr.15: 92094853-92094853 on Build GRCh38
rs57363993 Chr.15: 92094833-92094833 on Build GRCh38
rs1032993082 Chr.15: 92094997-92094997 on Build GRCh38
rs575451605 Chr.15: 92094827-92094827 on Build GRCh38
rs1001147768 Chr.15: 92094970-92094970 on Build GRCh38
rs368758974 Chr.15: 92094874-92094874 on Build GRCh38
rs778485345 Chr.15: 92094961-92094961 on Build GRCh38
rs111286820 Chr.15: 92094893-92094893 on Build GRCh38
rs371056448 Chr.15: 92094830-92094830 on Build GRCh38
rs765311316 Chr.15: 92094857-92094857 on Build GRCh38
rs956913139 Chr.15: 92095008-92095008 on Build GRCh38
rs544511825 Chr.15: 92095039-92095039 on Build GRCh38
rs775608978 Chr.15: 92095006-92095006 on Build GRCh38
rs753370739 Chr.15: 92095031-92095031 on Build GRCh38
rs759541723 Chr.15: 92094854-92094854 on Build GRCh38
rs199972220 Chr.15: 92094918-92094918 on Build GRCh38
rs767325626 Chr.15: 92094891-92094891 on Build GRCh38
rs372855100 Chr.15: 92094839-92094839 on Build GRCh38
rs2286355 Chr.15: 92094963-92094963 on Build GRCh38
rs1049434829 Chr.15: 92094852-92094852 on Build GRCh38
rs779641054 Chr.15: 92094898-92094898 on Build GRCh38
rs201929272 Chr.15: 92095014-92095014 on Build GRCh38
rs774235590 Chr.15: 92094834-92094834 on Build GRCh38
rs769719552 Chr.15: 92095003-92095003 on Build GRCh38
rs757754422 Chr.15: 92094887-92094887 on Build GRCh38
rs746970118 Chr.15: 92094979-92094979 on Build GRCh38
rs762991469 Chr.15: 92095012-92095012 on Build GRCh38
rs921629158 Chr.15: 92095032-92095032 on Build GRCh38
rs777032284 Chr.15: 92094876-92094876 on Build GRCh38
rs376741565 Chr.15: 92095020-92095020 on Build GRCh38
rs931941322 Chr.15: 92094845-92094845 on Build GRCh38
rs773198638 Chr.15: 92094843-92094843 on Build GRCh38
rs755629382 Chr.15: 92094896-92094896 on Build GRCh38
rs766839745 Chr.15: 92095025-92095025 on Build GRCh38

View Full Product Details