Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3370930-3371203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF195
Entrez Gene ID
7748
Gene Name
zinc finger protein 195
Gene Aliases
HRF1, ZNFP104
UniGene
Hs.386294
Gene Chromosome Location
Chr.11: 3357927-3379222 on Build GRCh38
Transcripts
NM_001130519.2 NM_001130520.2 NM_001242841.1 NM_001242842.1 NM_001242843.1 NM_001256823.1 NM_001256824.1 NM_001256825.1 NM_007152.4 NR_040083.1 NR_046381.1 NR_046382.1 XM_006718306.1 XM_011520350.2 XM_011520351.2 XM_011520352.2 XM_011520354.2 XM_017018260.1 XM_017018261.1 XM_017018262.1 XM_017018263.1 XM_017018264.1 XM_017018265.1 XM_017018266.1 XM_017018267.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4032500 c.216C>T p.D72D Chr.11: 3370985-3370985 on Build GRCh38
6485596 c.145A>G p.K49E Chr.11: 3371056-3371056 on Build GRCh38
3809138 c.195G>A p.V65V Chr.11: 3371006-3371006 on Build GRCh38
6828288 c.217G>A p.G73R Chr.11: 3370984-3370984 on Build GRCh38
6330124 c.187C>G p.R63G Chr.11: 3371026-3371026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372392415 Chr.11: 3371078-3371078 on Build GRCh38
rs375518869 Chr.11: 3370960-3370960 on Build GRCh38
rs2269734 Chr.11: 3371127-3371127 on Build GRCh38
rs776297846 Chr.11: 3371062-3371062 on Build GRCh38
rs547862465 Chr.11: 3371101-3371101 on Build GRCh38
rs766611036 Chr.11: 3370976-3370976 on Build GRCh38
rs79807062 Chr.11: 3371134-3371134 on Build GRCh38
rs550106470 Chr.11: 3371026-3371026 on Build GRCh38
rs2269735 Chr.11: 3371121-3371121 on Build GRCh38
rs764417930 Chr.11: 3370994-3370994 on Build GRCh38
rs368444165 Chr.11: 3370981-3370981 on Build GRCh38
rs183301949 Chr.11: 3370958-3370958 on Build GRCh38
rs772030493 Chr.11: 3371036-3371036 on Build GRCh38
rs764607061 Chr.11: 3370972-3370972 on Build GRCh38
rs774978578 Chr.11: 3371077-3371077 on Build GRCh38
rs1013825038 Chr.11: 3370993-3370993 on Build GRCh38
rs915554713 Chr.11: 3371132-3371132 on Build GRCh38
rs746786235 Chr.11: 3371053-3371053 on Build GRCh38
rs778995294 Chr.11: 3371025-3371025 on Build GRCh38
rs777476855 Chr.11: 3371043-3371043 on Build GRCh38
rs755047059 Chr.11: 3371004-3371004 on Build GRCh38
rs571847831 Chr.11: 3371074-3371074 on Build GRCh38
rs60703697 Chr.11: 3371083-3371083 on Build GRCh38
rs750795590 Chr.11: 3371088-3371088 on Build GRCh38
rs770521516 Chr.11: 3371058-3371058 on Build GRCh38
rs760299558 Chr.11: 3370959-3370959 on Build GRCh38
rs750790659 Chr.11: 3370984-3370984 on Build GRCh38
rs759004408 Chr.11: 3370966-3370966 on Build GRCh38
rs769226968 Chr.11: 3371076-3371076 on Build GRCh38
rs200059326 Chr.11: 3370963-3370963 on Build GRCh38
rs776858832 Chr.11: 3371010-3371010 on Build GRCh38
rs766536185 Chr.11: 3371117-3371117 on Build GRCh38
rs756343256 Chr.11: 3370985-3370985 on Build GRCh38
rs911193954 Chr.11: 3371142-3371142 on Build GRCh38
rs754005777 Chr.11: 3370997-3370997 on Build GRCh38

View Full Product Details