Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89098747-89099020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PM20D2
Entrez Gene ID
135293
Gene Name
peptidase M20 domain containing 2
Gene Aliases
ACY1L2, bA63L7.3
UniGene
Hs.356247
Gene Chromosome Location
Chr.6: 89096879-89165569 on Build GRCh38
Transcripts
XM_011535481.2 NM_080743.4 XM_006715348.2 XM_011535483.2 XM_011535484.1 XM_017010292.1 XM_017010293.1

Gene Symbol
SRSF12
Entrez Gene ID
135295
Gene Name
serine and arginine rich splicing factor 12
Gene Aliases
SFRS13B, SFRS19, SRrp35
UniGene
Hs.254414
Gene Chromosome Location
Chr.6: 89095957-89118081 on Build GRCh38
Transcripts
XM_011535481.2 NM_080743.4 XM_006715348.2 XM_011535483.2 XM_011535484.1 XM_017010292.1 XM_017010293.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3630448 c.584C>A p.S195* Chr.6: 89098780-89098780 on Build GRCh38
4827521 c.533G>A p.G178E Chr.6: 89098831-89098831 on Build GRCh38
1225455 c.470T>G p.L157* Chr.6: 89098894-89098894 on Build GRCh38
3748911 c.602A>C p.Q201P Chr.6: 89098762-89098762 on Build GRCh38
6301960 c.502A>G p.R168G Chr.6: 89098862-89098862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142638832 Chr.6: 89098975-89098975 on Build GRCh38
rs778295171 Chr.6: 89098880-89098880 on Build GRCh38
rs756560840 Chr.6: 89098866-89098866 on Build GRCh38
rs767177451 Chr.6: 89098968-89098968 on Build GRCh38
rs774197207 Chr.6: 89098962-89098962 on Build GRCh38
rs754200150 Chr.6: 89098885-89098885 on Build GRCh38
rs960934143 Chr.6: 89098817-89098817 on Build GRCh38
rs762879261 Chr.6: 89098829-89098829 on Build GRCh38
rs966413270 Chr.6: 89098971-89098971 on Build GRCh38
rs767921907 Chr.6: 89098976-89098976 on Build GRCh38
rs779351857 Chr.6: 89098905-89098905 on Build GRCh38
rs746010685 Chr.6: 89098916-89098916 on Build GRCh38
rs374369058 Chr.6: 89098934-89098934 on Build GRCh38
rs772370071 Chr.6: 89098923-89098923 on Build GRCh38
rs775716564 Chr.6: 89098791-89098791 on Build GRCh38
rs745951507 Chr.6: 89098780-89098780 on Build GRCh38
rs1018358315 Chr.6: 89098906-89098906 on Build GRCh38
rs771989086 Chr.6: 89098789-89098789 on Build GRCh38
rs201226352 Chr.6: 89098771-89098771 on Build GRCh38
rs560814640 Chr.6: 89098861-89098861 on Build GRCh38
rs766050040 Chr.6: 89098839-89098839 on Build GRCh38
rs773348792 Chr.6: 89098935-89098935 on Build GRCh38
rs761804002 Chr.6: 89098800-89098800 on Build GRCh38
rs766868282 Chr.6: 89098853-89098853 on Build GRCh38
rs192903351 Chr.6: 89098816-89098816 on Build GRCh38
rs780234250 Chr.6: 89098933-89098933 on Build GRCh38
rs759066593 Chr.6: 89098965-89098965 on Build GRCh38
rs760248765 Chr.6: 89098989-89098989 on Build GRCh38
rs771155895 Chr.6: 89098770-89098770 on Build GRCh38
rs759236309 Chr.6: 89098852-89098852 on Build GRCh38
rs200555327 Chr.6: 89098939-89098939 on Build GRCh38
rs199535774 Chr.6: 89098927-89098927 on Build GRCh38
rs201200624 Chr.6: 89098828-89098828 on Build GRCh38
rs17851929 Chr.6: 89098919-89098919 on Build GRCh38
rs745603920 Chr.6: 89098840-89098840 on Build GRCh38
rs370679383 Chr.6: 89098821-89098821 on Build GRCh38
rs769875813 Chr.6: 89098823-89098823 on Build GRCh38
rs770572873 Chr.6: 89098950-89098950 on Build GRCh38
rs757624112 Chr.6: 89098886-89098886 on Build GRCh38

View Full Product Details