Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 85012717-85012942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA3D
Entrez Gene ID
223117
Gene Name
semaphorin 3D
Gene Aliases
Sema-Z2, coll-2
UniGene
Hs.201340
Gene Chromosome Location
Chr.7: 84995192-85187176 on Build GRCh38
Transcripts
NM_152754.2 XM_011515960.1 XM_011515961.2 XM_017011873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3950872 c.1765G>T p.D589Y Chr.7: 85012785-85012785 on Build GRCh38
461527 c.1734C>T p.G578G Chr.7: 85012816-85012816 on Build GRCh38
5750340 c.1739C>A p.P580Q Chr.7: 85012811-85012811 on Build GRCh38
1187345 c.1754G>T p.W585L Chr.7: 85012796-85012796 on Build GRCh38
6040274 c.1768+1G>A p.? Chr.7: 85012781-85012781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748974910 Chr.7: 85012860-85012860 on Build GRCh38
rs1044383735 Chr.7: 85012789-85012789 on Build GRCh38
rs576069112 Chr.7: 85012763-85012763 on Build GRCh38
rs754910513 Chr.7: 85012836-85012836 on Build GRCh38
rs141821755 Chr.7: 85012830-85012830 on Build GRCh38
rs753111661 Chr.7: 85012828-85012828 on Build GRCh38
rs779974681 Chr.7: 85012757-85012757 on Build GRCh38
rs767894621 Chr.7: 85012821-85012821 on Build GRCh38
rs543079856 Chr.7: 85012866-85012866 on Build GRCh38
rs1013587008 Chr.7: 85012754-85012754 on Build GRCh38
rs182694281 Chr.7: 85012776-85012776 on Build GRCh38
rs781016117 Chr.7: 85012839-85012839 on Build GRCh38
rs375254494 Chr.7: 85012815-85012815 on Build GRCh38
rs780878639 Chr.7: 85012774-85012774 on Build GRCh38
rs367647437 Chr.7: 85012769-85012769 on Build GRCh38
rs775829437 Chr.7: 85012884-85012884 on Build GRCh38
rs199662406 Chr.7: 85012882-85012882 on Build GRCh38
rs200711770 Chr.7: 85012850-85012850 on Build GRCh38
rs768448304 Chr.7: 85012773-85012773 on Build GRCh38
rs781759000 Chr.7: 85012788-85012788 on Build GRCh38
rs545463042 Chr.7: 85012791-85012791 on Build GRCh38
rs760962525 Chr.7: 85012892-85012892 on Build GRCh38
rs775126266 Chr.7: 85012747-85012747 on Build GRCh38
rs553108838 Chr.7: 85012771-85012771 on Build GRCh38
rs1048262415 Chr.7: 85012913-85012913 on Build GRCh38
rs150646719 Chr.7: 85012835-85012835 on Build GRCh38
rs776783624 Chr.7: 85012895-85012895 on Build GRCh38
rs372151810 Chr.7: 85012804-85012804 on Build GRCh38
rs1057519374 Chr.7: 85012812-85012812 on Build GRCh38
rs762385798 Chr.7: 85012886-85012890 on Build GRCh38
rs757269487 Chr.7: 85012831-85012831 on Build GRCh38
rs748011911 Chr.7: 85012846-85012846 on Build GRCh38
rs532983498 Chr.7: 85012893-85012893 on Build GRCh38
rs1032514707 Chr.7: 85012792-85012792 on Build GRCh38
rs762614894 Chr.7: 85012795-85012795 on Build GRCh38
rs1002762802 Chr.7: 85012813-85012813 on Build GRCh38
rs575976520 Chr.7: 85012851-85012851 on Build GRCh38
rs753977126 Chr.7: 85012756-85012756 on Build GRCh38
rs748562150 Chr.7: 85012816-85012816 on Build GRCh38
rs746366972 Chr.7: 85012873-85012873 on Build GRCh38

View Full Product Details