Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42310638-42310911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GANC
Entrez Gene ID
2595
Gene Name
glucosidase alpha, neutral C
Gene Aliases
-
UniGene
Hs.730806
Gene Chromosome Location
Chr.15: 42273658-42353666 on Build GRCh38
Transcripts
NM_001301409.1 NM_198141.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6898660 c.928G>A p.V310I Chr.15: 42310717-42310717 on Build GRCh38
6463883 c.907A>T p.T303S Chr.15: 42310696-42310696 on Build GRCh38
1749133 c.956G>A p.R319H Chr.15: 42310745-42310745 on Build GRCh38
1290382 c.1036A>T p.K346* Chr.15: 42310825-42310825 on Build GRCh38
700591 c.905A>T p.Y302F Chr.15: 42310694-42310694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764397658 Chr.15: 42310707-42310707 on Build GRCh38
rs757653747 Chr.15: 42310748-42310748 on Build GRCh38
rs2412694 Chr.15: 42310885-42310885 on Build GRCh38
rs139559678 Chr.15: 42310680-42310680 on Build GRCh38
rs770690162 Chr.15: 42310751-42310751 on Build GRCh38
rs746171994 Chr.15: 42310799-42310799 on Build GRCh38
rs111569626 Chr.15: 42310698-42310698 on Build GRCh38
rs1040674935 Chr.15: 42310697-42310697 on Build GRCh38
rs772649624 Chr.15: 42310800-42310800 on Build GRCh38
rs34620840 Chr.15: 42310688-42310689 on Build GRCh38
rs930461810 Chr.15: 42310851-42310851 on Build GRCh38
rs200635553 Chr.15: 42310856-42310856 on Build GRCh38
rs749704363 Chr.15: 42310662-42310662 on Build GRCh38
rs776977110 Chr.15: 42310666-42310666 on Build GRCh38
rs975211410 Chr.15: 42310835-42310835 on Build GRCh38
rs911861951 Chr.15: 42310786-42310786 on Build GRCh38
rs762015762 Chr.15: 42310844-42310844 on Build GRCh38
rs987461905 Chr.15: 42310757-42310757 on Build GRCh38
rs763396631 Chr.15: 42310862-42310862 on Build GRCh38
rs376380569 Chr.15: 42310668-42310668 on Build GRCh38
rs766212135 Chr.15: 42310724-42310724 on Build GRCh38
rs143247407 Chr.15: 42310695-42310695 on Build GRCh38
rs779329647 Chr.15: 42310751-42310751 on Build GRCh38
rs772450391 Chr.15: 42310687-42310687 on Build GRCh38
rs201403785 Chr.15: 42310781-42310781 on Build GRCh38
rs754914062 Chr.15: 42310737-42310737 on Build GRCh38
rs769764283 Chr.15: 42310854-42310854 on Build GRCh38
rs768659539 Chr.15: 42310829-42310829 on Build GRCh38
rs551903460 Chr.15: 42310860-42310860 on Build GRCh38
rs141204390 Chr.15: 42310740-42310740 on Build GRCh38
rs751351033 Chr.15: 42310727-42310727 on Build GRCh38
rs761992646 Chr.15: 42310723-42310723 on Build GRCh38
rs964711791 Chr.15: 42310826-42310826 on Build GRCh38
rs751369094 Chr.15: 42310849-42310849 on Build GRCh38
rs760610447 Chr.15: 42310705-42310705 on Build GRCh38
rs748993441 Chr.15: 42310748-42310748 on Build GRCh38
rs939235459 Chr.15: 42310884-42310884 on Build GRCh38
rs956315688 Chr.15: 42310755-42310755 on Build GRCh38
rs377160213 Chr.15: 42310745-42310745 on Build GRCh38
rs1022748199 Chr.15: 42310831-42310831 on Build GRCh38
rs776670979 Chr.15: 42310832-42310832 on Build GRCh38
rs756978278 Chr.15: 42310886-42310886 on Build GRCh38
rs774838868 Chr.15: 42310678-42310678 on Build GRCh38
rs1012370094 Chr.15: 42310670-42310670 on Build GRCh38
rs763750972 Chr.15: 42310875-42310875 on Build GRCh38
rs746390371 Chr.15: 42310781-42310781 on Build GRCh38
rs969261502 Chr.15: 42310708-42310708 on Build GRCh38
rs752273058 Chr.15: 42310744-42310744 on Build GRCh38
rs767519840 Chr.15: 42310857-42310857 on Build GRCh38
rs754020623 Chr.15: 42310722-42310722 on Build GRCh38
rs756053987 Chr.15: 42310871-42310871 on Build GRCh38
rs760841430 Chr.15: 42310818-42310818 on Build GRCh38
rs150733805 Chr.15: 42310812-42310812 on Build GRCh38

View Full Product Details