Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5696587-5696828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LONP1
Entrez Gene ID
9361
Gene Name
lon peptidase 1, mitochondrial
Gene Aliases
CODASS, LON, LONP, LonHS, PIM1, PRSS15, hLON
UniGene
Hs.350265
Gene Chromosome Location
Chr.19: 5691834-5720452 on Build GRCh38
Transcripts
NM_001276479.1 NM_001276480.1 NM_004793.3 NR_076392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5468444 c.1773+4C>T p.? Chr.19: 5696666-5696666 on Build GRCh38
7332911 c.1764C>T p.L588L Chr.19: 5696679-5696679 on Build GRCh38
5632559 c.1727G>A p.C576Y Chr.19: 5696716-5696716 on Build GRCh38
7348151 c.1702G>A p.A568T Chr.19: 5696741-5696741 on Build GRCh38
6003467 c.1770C>T p.D590D Chr.19: 5696673-5696673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003466635 Chr.19: 5696656-5696657 on Build GRCh38
rs770066551 Chr.19: 5696717-5696717 on Build GRCh38
rs768211684 Chr.19: 5696611-5696612 on Build GRCh38
rs78785368 Chr.19: 5696609-5696609 on Build GRCh38
rs746194260 Chr.19: 5696641-5696642 on Build GRCh38
rs372931156 Chr.19: 5696639-5696640 on Build GRCh38
rs138649872 Chr.19: 5696697-5696697 on Build GRCh38
rs777108243 Chr.19: 5696640-5696641 on Build GRCh38
rs774604129 Chr.19: 5696799-5696799 on Build GRCh38
rs185333714 Chr.19: 5696640-5696640 on Build GRCh38
rs576425895 Chr.19: 5696772-5696772 on Build GRCh38
rs748250841 Chr.19: 5696709-5696709 on Build GRCh38
rs371952742 Chr.19: 5696626-5696626 on Build GRCh38
rs746321328 Chr.19: 5696659-5696659 on Build GRCh38
rs375197253 Chr.19: 5696764-5696764 on Build GRCh38
rs766960869 Chr.19: 5696807-5696809 on Build GRCh38
rs747368479 Chr.19: 5696664-5696664 on Build GRCh38
rs761635410 Chr.19: 5696807-5696807 on Build GRCh38
rs147317335 Chr.19: 5696622-5696622 on Build GRCh38
rs776493049 Chr.19: 5696804-5696804 on Build GRCh38
rs752552069 Chr.19: 5696641-5696641 on Build GRCh38
rs769047081 Chr.19: 5696665-5696665 on Build GRCh38
rs552289191 Chr.19: 5696806-5696806 on Build GRCh38
rs200985875 Chr.19: 5696661-5696661 on Build GRCh38
rs1003183270 Chr.19: 5696788-5696788 on Build GRCh38
rs112238400 Chr.19: 5696773-5696773 on Build GRCh38
rs778508442 Chr.19: 5696655-5696655 on Build GRCh38
rs11551018 Chr.19: 5696676-5696676 on Build GRCh38
rs372845163 Chr.19: 5696784-5696784 on Build GRCh38
rs756969382 Chr.19: 5696690-5696690 on Build GRCh38
rs546406418 Chr.19: 5696790-5696790 on Build GRCh38
rs775776260 Chr.19: 5696662-5696662 on Build GRCh38
rs150208648 Chr.19: 5696727-5696727 on Build GRCh38
rs764931850 Chr.19: 5696750-5696750 on Build GRCh38
rs767351728 Chr.19: 5696639-5696639 on Build GRCh38
rs138529751 Chr.19: 5696748-5696748 on Build GRCh38
rs144817438 Chr.19: 5696785-5696785 on Build GRCh38
rs747561131 Chr.19: 5696699-5696699 on Build GRCh38
rs750112967 Chr.19: 5696751-5696751 on Build GRCh38
rs934344386 Chr.19: 5696606-5696606 on Build GRCh38
rs370718882 Chr.19: 5696666-5696666 on Build GRCh38
rs994420325 Chr.19: 5696795-5696795 on Build GRCh38
rs892884949 Chr.19: 5696610-5696610 on Build GRCh38
rs760821454 Chr.19: 5696625-5696625 on Build GRCh38
rs369256803 Chr.19: 5696802-5696802 on Build GRCh38
rs368483569 Chr.19: 5696783-5696783 on Build GRCh38
rs188167373 Chr.19: 5696797-5696797 on Build GRCh38
rs181481775 Chr.19: 5696635-5696635 on Build GRCh38
rs200413275 Chr.19: 5696803-5696803 on Build GRCh38
rs757878491 Chr.19: 5696757-5696757 on Build GRCh38
rs775864308 Chr.19: 5696624-5696624 on Build GRCh38
rs772860258 Chr.19: 5696810-5696812 on Build GRCh38
rs201834225 Chr.19: 5696636-5696636 on Build GRCh38
rs759487151 Chr.19: 5696677-5696677 on Build GRCh38
rs374023976 Chr.19: 5696673-5696673 on Build GRCh38
rs376466825 Chr.19: 5696651-5696651 on Build GRCh38
rs1035965117 Chr.19: 5696646-5696646 on Build GRCh38
rs200136098 Chr.19: 5696733-5696733 on Build GRCh38
rs763761431 Chr.19: 5696739-5696739 on Build GRCh38
rs761338260 Chr.19: 5696778-5696779 on Build GRCh38
rs368178461 Chr.19: 5696678-5696678 on Build GRCh38
rs199854352 Chr.19: 5696627-5696627 on Build GRCh38
rs773361968 Chr.19: 5696718-5696718 on Build GRCh38
rs763122032 Chr.19: 5696724-5696724 on Build GRCh38
rs755512692 Chr.19: 5696776-5696776 on Build GRCh38
rs58283603 Chr.19: 5696809-5696809 on Build GRCh38
rs761766740 Chr.19: 5696629-5696629 on Build GRCh38
rs371125440 Chr.19: 5696652-5696652 on Build GRCh38
rs373671256 Chr.19: 5696653-5696653 on Build GRCh38
rs775939635 Chr.19: 5696779-5696779 on Build GRCh38
rs140317309 Chr.19: 5696698-5696698 on Build GRCh38
rs138807867 Chr.19: 5696736-5696736 on Build GRCh38
rs147468253 Chr.19: 5696742-5696742 on Build GRCh38
rs149349963 Chr.19: 5696738-5696738 on Build GRCh38
rs570323541 Chr.19: 5696756-5696756 on Build GRCh38
rs200147790 Chr.19: 5696707-5696707 on Build GRCh38
rs753388578 Chr.19: 5696687-5696687 on Build GRCh38
rs778155266 Chr.19: 5696630-5696631 on Build GRCh38
rs768956788 Chr.19: 5696701-5696701 on Build GRCh38
rs776972476 Chr.19: 5696685-5696685 on Build GRCh38
rs756034543 Chr.19: 5696647-5696647 on Build GRCh38
rs770370766 Chr.19: 5696765-5696765 on Build GRCh38
rs3840916 Chr.19: 5696635-5696636 on Build GRCh38

View Full Product Details