Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206901891-206902160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL24
Entrez Gene ID
11009
Gene Name
interleukin 24
Gene Aliases
C49A, FISP, IL10B, MDA7, MOB5, ST16
UniGene
Hs.58831
Gene Chromosome Location
Chr.1: 206897404-206904139 on Build GRCh38
Transcripts
NM_001185156.1 NM_001185157.1 NM_001185158.1 NM_006850.3 XM_011509101.2 XM_017000121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
117430 c.508T>A p.F170I Chr.1: 206902043-206902043 on Build GRCh38
6685254 c.495T>C p.S165S Chr.1: 206902030-206902030 on Build GRCh38
903021 c.522G>A p.R174R Chr.1: 206902057-206902057 on Build GRCh38
5992481 c.521G>A p.R174Q Chr.1: 206902056-206902056 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs147989968 Chr.1: 206902055-206902055 on Build GRCh38
rs753278690 Chr.1: 206901961-206901961 on Build GRCh38
rs764528690 Chr.1: 206901972-206901972 on Build GRCh38
rs766443483 Chr.1: 206901949-206901949 on Build GRCh38
rs765771975 Chr.1: 206901959-206901959 on Build GRCh38
rs764357673 Chr.1: 206902054-206902054 on Build GRCh38
rs753715416 Chr.1: 206902083-206902083 on Build GRCh38
rs535005816 Chr.1: 206902139-206902139 on Build GRCh38
rs776658873 Chr.1: 206902031-206902031 on Build GRCh38
rs1015047238 Chr.1: 206902100-206902100 on Build GRCh38
rs371070306 Chr.1: 206902007-206902007 on Build GRCh38
rs369059255 Chr.1: 206901958-206901958 on Build GRCh38
rs200334512 Chr.1: 206902095-206902095 on Build GRCh38
rs775803587 Chr.1: 206902041-206902041 on Build GRCh38
rs1049881754 Chr.1: 206901927-206901927 on Build GRCh38
rs755002501 Chr.1: 206902085-206902085 on Build GRCh38
rs768654212 Chr.1: 206901950-206901950 on Build GRCh38
rs771193700 Chr.1: 206902030-206902030 on Build GRCh38
rs976225422 Chr.1: 206902140-206902140 on Build GRCh38
rs79685336 Chr.1: 206902043-206902043 on Build GRCh38
rs756998158 Chr.1: 206901982-206901982 on Build GRCh38
rs780833295 Chr.1: 206901983-206901983 on Build GRCh38
rs1040185766 Chr.1: 206902020-206902020 on Build GRCh38
rs779620585 Chr.1: 206902070-206902070 on Build GRCh38
rs781341202 Chr.1: 206902105-206902105 on Build GRCh38
rs200977694 Chr.1: 206901988-206901988 on Build GRCh38
rs755631667 Chr.1: 206902066-206902066 on Build GRCh38
rs377755475 Chr.1: 206902040-206902041 on Build GRCh38
rs748012828 Chr.1: 206902101-206902101 on Build GRCh38
rs202138928 Chr.1: 206901955-206901955 on Build GRCh38
rs759570553 Chr.1: 206902037-206902037 on Build GRCh38
rs539103349 Chr.1: 206902056-206902056 on Build GRCh38
rs376493626 Chr.1: 206901993-206901993 on Build GRCh38
rs751102642 Chr.1: 206901979-206901979 on Build GRCh38
rs146604458 Chr.1: 206902040-206902040 on Build GRCh38
rs889517289 Chr.1: 206901938-206901938 on Build GRCh38
rs372509817 Chr.1: 206901968-206901968 on Build GRCh38
rs185504632 Chr.1: 206902102-206902102 on Build GRCh38
rs777408537 Chr.1: 206902117-206902117 on Build GRCh38
rs114904491 Chr.1: 206902063-206902063 on Build GRCh38
rs778835664 Chr.1: 206902023-206902023 on Build GRCh38
rs763516781 Chr.1: 206902049-206902049 on Build GRCh38
rs368977081 Chr.1: 206902094-206902094 on Build GRCh38
rs747187073 Chr.1: 206902027-206902027 on Build GRCh38
rs946445418 Chr.1: 206901944-206901944 on Build GRCh38
rs768269238 Chr.1: 206902011-206902011 on Build GRCh38
rs376804393 Chr.1: 206901946-206901946 on Build GRCh38

View Full Product Details