Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144464101-144464366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYHR1
Entrez Gene ID
50626
Gene Name
cysteine and histidine rich 1
Gene Aliases
CHRP
UniGene
Hs.459379
Gene Chromosome Location
Chr.8: 144449582-144465692 on Build GRCh38
Transcripts
NM_001129888.1 NM_032687.3 NM_138496.1 XM_006716560.3 XM_006716561.3 XM_017013463.1 XM_017013464.1 XM_017013465.1 XM_017013466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
145293 c.422G>C p.W141S Chr.8: 144464284-144464284 on Build GRCh38
4977123 c.469G>A p.E157K Chr.8: 144464237-144464237 on Build GRCh38
1569135 c.544C>T p.R182W Chr.8: 144464162-144464162 on Build GRCh38
5430204 c.446C>G p.A149G Chr.8: 144464260-144464260 on Build GRCh38
1455657 c.430delG p.A144fs*9 Chr.8: 144464276-144464276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773213525 Chr.8: 144464349-144464349 on Build GRCh38
rs370071758 Chr.8: 144464136-144464136 on Build GRCh38
rs770367627 Chr.8: 144464199-144464199 on Build GRCh38
rs762099793 Chr.8: 144464276-144464276 on Build GRCh38
rs371644139 Chr.8: 144464324-144464324 on Build GRCh38
rs143628277 Chr.8: 144464283-144464283 on Build GRCh38
rs543509475 Chr.8: 144464162-144464162 on Build GRCh38
rs547376015 Chr.8: 144464275-144464276 on Build GRCh38
rs764328897 Chr.8: 144464298-144464298 on Build GRCh38
rs757794751 Chr.8: 144464267-144464267 on Build GRCh38
rs148681176 Chr.8: 144464304-144464304 on Build GRCh38
rs764367953 Chr.8: 144464233-144464235 on Build GRCh38
rs368285970 Chr.8: 144464311-144464311 on Build GRCh38
rs1018944792 Chr.8: 144464159-144464159 on Build GRCh38
rs375882093 Chr.8: 144464292-144464292 on Build GRCh38
rs77204173 Chr.8: 144464214-144464214 on Build GRCh38
rs749590903 Chr.8: 144464120-144464120 on Build GRCh38
rs761863174 Chr.8: 144464232-144464232 on Build GRCh38
rs762642971 Chr.8: 144464215-144464215 on Build GRCh38
rs778756479 Chr.8: 144464329-144464329 on Build GRCh38
rs764965148 Chr.8: 144464153-144464153 on Build GRCh38
rs578183711 Chr.8: 144464198-144464198 on Build GRCh38
rs753025595 Chr.8: 144464253-144464253 on Build GRCh38
rs758621338 Chr.8: 144464256-144464256 on Build GRCh38
rs779440226 Chr.8: 144464127-144464127 on Build GRCh38
rs775184067 Chr.8: 144464154-144464154 on Build GRCh38
rs751572780 Chr.8: 144464158-144464158 on Build GRCh38
rs774091305 Chr.8: 144464224-144464224 on Build GRCh38
rs762849718 Chr.8: 144464155-144464155 on Build GRCh38
rs919087758 Chr.8: 144464184-144464184 on Build GRCh38
rs772055597 Chr.8: 144464340-144464340 on Build GRCh38
rs199539352 Chr.8: 144464302-144464302 on Build GRCh38
rs748648310 Chr.8: 144464133-144464133 on Build GRCh38
rs768898712 Chr.8: 144464160-144464160 on Build GRCh38
rs915862154 Chr.8: 144464337-144464337 on Build GRCh38
rs764944855 Chr.8: 144464138-144464138 on Build GRCh38
rs778210137 Chr.8: 144464259-144464259 on Build GRCh38
rs201021717 Chr.8: 144464194-144464194 on Build GRCh38
rs781697158 Chr.8: 144464268-144464268 on Build GRCh38
rs748438702 Chr.8: 144464185-144464185 on Build GRCh38
rs142152732 Chr.8: 144464284-144464284 on Build GRCh38
rs966082253 Chr.8: 144464167-144464167 on Build GRCh38
rs146960526 Chr.8: 144464281-144464281 on Build GRCh38
rs776952885 Chr.8: 144464191-144464191 on Build GRCh38
rs763917343 Chr.8: 144464156-144464156 on Build GRCh38
rs948382010 Chr.8: 144464234-144464234 on Build GRCh38
rs762990071 Chr.8: 144464216-144464217 on Build GRCh38
rs772426239 Chr.8: 144464134-144464134 on Build GRCh38
rs766525030 Chr.8: 144464240-144464240 on Build GRCh38
rs149259609 Chr.8: 144464280-144464280 on Build GRCh38
rs752016150 Chr.8: 144464262-144464262 on Build GRCh38
rs139843668 Chr.8: 144464282-144464282 on Build GRCh38
rs766572350 Chr.8: 144464164-144464164 on Build GRCh38
rs759737579 Chr.8: 144464244-144464244 on Build GRCh38
rs759681184 Chr.8: 144464285-144464285 on Build GRCh38
rs747942669 Chr.8: 144464332-144464332 on Build GRCh38
rs745363210 Chr.8: 144464276-144464276 on Build GRCh38
rs763256167 Chr.8: 144464296-144464296 on Build GRCh38
rs199972231 Chr.8: 144464202-144464202 on Build GRCh38
rs776196363 Chr.8: 144464145-144464145 on Build GRCh38
rs756644723 Chr.8: 144464313-144464316 on Build GRCh38
rs759202673 Chr.8: 144464152-144464152 on Build GRCh38
rs373353154 Chr.8: 144464166-144464166 on Build GRCh38
rs761161002 Chr.8: 144464144-144464144 on Build GRCh38
rs374378728 Chr.8: 144464309-144464309 on Build GRCh38
rs773814209 Chr.8: 144464135-144464135 on Build GRCh38
rs750926515 Chr.8: 144464284-144464285 on Build GRCh38
rs750123440 Chr.8: 144464170-144464170 on Build GRCh38
rs765359523 Chr.8: 144464289-144464289 on Build GRCh38
rs560617378 Chr.8: 144464205-144464205 on Build GRCh38
rs754062946 Chr.8: 144464168-144464168 on Build GRCh38
rs775498418 Chr.8: 144464133-144464134 on Build GRCh38
rs776144196 Chr.8: 144464201-144464201 on Build GRCh38
rs750996356 Chr.8: 144464310-144464310 on Build GRCh38
rs753937256 Chr.8: 144464242-144464242 on Build GRCh38
rs779632554 Chr.8: 144464321-144464321 on Build GRCh38
rs898772897 Chr.8: 144464331-144464331 on Build GRCh38
rs769405048 Chr.8: 144464279-144464279 on Build GRCh38
rs1045415312 Chr.8: 144464235-144464235 on Build GRCh38
rs765360904 Chr.8: 144464245-144464245 on Build GRCh38
rs377084522 Chr.8: 144464124-144464124 on Build GRCh38

View Full Product Details