Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 183492022-183492253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL6
Entrez Gene ID
89857
Gene Name
kelch like family member 6
Gene Aliases
-
UniGene
Hs.659147
Gene Chromosome Location
Chr.3: 183487531-183555712 on Build GRCh38
Transcripts
NM_130446.2 XM_011513273.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5484609 c.1751A>G p.Q584R Chr.3: 183492042-183492042 on Build GRCh38
6696877 c.1740C>T p.D580D Chr.3: 183492053-183492053 on Build GRCh38
1658914 c.1744G>A p.E582K Chr.3: 183492049-183492049 on Build GRCh38
6696881 c.1743delC p.E582fs*5 Chr.3: 183492050-183492050 on Build GRCh38
6696882 c.1654G>A p.G552S Chr.3: 183492139-183492139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751035968 Chr.3: 183492193-183492193 on Build GRCh38
rs376538225 Chr.3: 183492057-183492057 on Build GRCh38
rs760709738 Chr.3: 183492130-183492130 on Build GRCh38
rs1012832935 Chr.3: 183492051-183492051 on Build GRCh38
rs770057603 Chr.3: 183492208-183492208 on Build GRCh38
rs551153731 Chr.3: 183492115-183492115 on Build GRCh38
rs879612900 Chr.3: 183492067-183492069 on Build GRCh38
rs779586577 Chr.3: 183492180-183492180 on Build GRCh38
rs139938005 Chr.3: 183492098-183492098 on Build GRCh38
rs932157964 Chr.3: 183492194-183492194 on Build GRCh38
rs748455295 Chr.3: 183492104-183492104 on Build GRCh38
rs199528524 Chr.3: 183492212-183492212 on Build GRCh38
rs958836337 Chr.3: 183492043-183492043 on Build GRCh38
rs879371624 Chr.3: 183492071-183492079 on Build GRCh38
rs984794444 Chr.3: 183492074-183492074 on Build GRCh38
rs534097684 Chr.3: 183492155-183492155 on Build GRCh38
rs868826583 Chr.3: 183492139-183492139 on Build GRCh38
rs771158305 Chr.3: 183492122-183492122 on Build GRCh38
rs765294798 Chr.3: 183492136-183492137 on Build GRCh38
rs373496199 Chr.3: 183492050-183492050 on Build GRCh38
rs267599702 Chr.3: 183492200-183492200 on Build GRCh38
rs763935771 Chr.3: 183492133-183492133 on Build GRCh38
rs150096820 Chr.3: 183492209-183492209 on Build GRCh38
rs761531481 Chr.3: 183492140-183492140 on Build GRCh38
rs781535635 Chr.3: 183492201-183492201 on Build GRCh38
rs770111088 Chr.3: 183492113-183492113 on Build GRCh38
rs749902846 Chr.3: 183492233-183492233 on Build GRCh38
rs764948740 Chr.3: 183492146-183492146 on Build GRCh38
rs763201287 Chr.3: 183492118-183492118 on Build GRCh38
rs566509636 Chr.3: 183492128-183492128 on Build GRCh38
rs757004264 Chr.3: 183492045-183492045 on Build GRCh38
rs200447774 Chr.3: 183492205-183492205 on Build GRCh38
rs757992910 Chr.3: 183492177-183492177 on Build GRCh38
rs769019214 Chr.3: 183492092-183492092 on Build GRCh38
rs753728391 Chr.3: 183492134-183492134 on Build GRCh38
rs1021590457 Chr.3: 183492089-183492089 on Build GRCh38
rs376772219 Chr.3: 183492173-183492173 on Build GRCh38
rs779494995 Chr.3: 183492055-183492055 on Build GRCh38

View Full Product Details