Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49525390-49525651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGRT
Entrez Gene ID
2217
Gene Name
Fc fragment of IgG receptor and transporter
Gene Aliases
FCRN, alpha-chain
UniGene
Hs.111903
Gene Chromosome Location
Chr.19: 49512279-49526428 on Build GRCh38
Transcripts
NM_001136019.2 NM_004107.4 XM_005258657.1 XM_011526627.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6332347 c.887C>T p.S296F Chr.19: 49525472-49525472 on Build GRCh38
3672974 c.877C>G p.P293A Chr.19: 49525462-49525462 on Build GRCh38
999317 c.938C>T p.A313V Chr.19: 49525523-49525523 on Build GRCh38
3692960 c.916G>A p.G306S Chr.19: 49525501-49525501 on Build GRCh38
6662774 c.875C>A p.S292Y Chr.19: 49525460-49525460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758129958 Chr.19: 49525586-49525586 on Build GRCh38
rs758739321 Chr.19: 49525418-49525418 on Build GRCh38
rs750783192 Chr.19: 49525413-49525413 on Build GRCh38
rs371610444 Chr.19: 49525428-49525428 on Build GRCh38
rs982258593 Chr.19: 49525627-49525627 on Build GRCh38
rs780831224 Chr.19: 49525431-49525431 on Build GRCh38
rs773899345 Chr.19: 49525557-49525559 on Build GRCh38
rs961720168 Chr.19: 49525511-49525511 on Build GRCh38
rs752078473 Chr.19: 49525622-49525622 on Build GRCh38
rs757155105 Chr.19: 49525579-49525579 on Build GRCh38
rs1048767818 Chr.19: 49525617-49525618 on Build GRCh38
rs778783701 Chr.19: 49525581-49525581 on Build GRCh38
rs748397054 Chr.19: 49525598-49525598 on Build GRCh38
rs769845543 Chr.19: 49525530-49525530 on Build GRCh38
rs552700082 Chr.19: 49525495-49525495 on Build GRCh38
rs144900355 Chr.19: 49525501-49525501 on Build GRCh38
rs200666118 Chr.19: 49525523-49525523 on Build GRCh38
rs34807374 Chr.19: 49525518-49525518 on Build GRCh38
rs760174728 Chr.19: 49525576-49525576 on Build GRCh38
rs771467179 Chr.19: 49525542-49525542 on Build GRCh38
rs749795129 Chr.19: 49525617-49525617 on Build GRCh38
rs757058501 Chr.19: 49525506-49525506 on Build GRCh38
rs575406297 Chr.19: 49525585-49525585 on Build GRCh38
rs973257707 Chr.19: 49525514-49525514 on Build GRCh38
rs748189145 Chr.19: 49525528-49525528 on Build GRCh38
rs929673388 Chr.19: 49525604-49525604 on Build GRCh38
rs763649919 Chr.19: 49525577-49525577 on Build GRCh38
rs199588435 Chr.19: 49525505-49525505 on Build GRCh38
rs141001122 Chr.19: 49525483-49525483 on Build GRCh38
rs771593747 Chr.19: 49525623-49525623 on Build GRCh38
rs775759872 Chr.19: 49525469-49525469 on Build GRCh38
rs752280594 Chr.19: 49525498-49525498 on Build GRCh38
rs767079527 Chr.19: 49525612-49525613 on Build GRCh38
rs927498603 Chr.19: 49525576-49525576 on Build GRCh38
rs902540996 Chr.19: 49525467-49525467 on Build GRCh38
rs538290471 Chr.19: 49525500-49525500 on Build GRCh38
rs752330495 Chr.19: 49525572-49525572 on Build GRCh38
rs777720517 Chr.19: 49525454-49525454 on Build GRCh38
rs763500395 Chr.19: 49525537-49525537 on Build GRCh38
rs111846329 Chr.19: 49525474-49525474 on Build GRCh38
rs181182779 Chr.19: 49525596-49525596 on Build GRCh38
rs148496736 Chr.19: 49525552-49525552 on Build GRCh38
rs778656931 Chr.19: 49525517-49525517 on Build GRCh38
rs745717269 Chr.19: 49525554-49525554 on Build GRCh38
rs140449664 Chr.19: 49525461-49525461 on Build GRCh38
rs761385658 Chr.19: 49525575-49525576 on Build GRCh38
rs879009556 Chr.19: 49525455-49525455 on Build GRCh38
rs373170275 Chr.19: 49525601-49525601 on Build GRCh38
rs781341160 Chr.19: 49525524-49525524 on Build GRCh38
rs765674997 Chr.19: 49525412-49525412 on Build GRCh38
rs199893057 Chr.19: 49525482-49525482 on Build GRCh38
rs753770315 Chr.19: 49525578-49525578 on Build GRCh38
rs34522905 Chr.19: 49525477-49525477 on Build GRCh38
rs201536964 Chr.19: 49525497-49525497 on Build GRCh38
rs777883975 Chr.19: 49525610-49525610 on Build GRCh38
rs754598494 Chr.19: 49525430-49525430 on Build GRCh38
rs773658267 Chr.19: 49525535-49525535 on Build GRCh38
rs766845001 Chr.19: 49525487-49525487 on Build GRCh38
rs770697747 Chr.19: 49525466-49525466 on Build GRCh38
rs374753638 Chr.19: 49525494-49525494 on Build GRCh38
rs752280951 Chr.19: 49525442-49525442 on Build GRCh38
rs748142914 Chr.19: 49525441-49525441 on Build GRCh38
rs142797093 Chr.19: 49525571-49525571 on Build GRCh38
rs893945588 Chr.19: 49525479-49525479 on Build GRCh38
rs145875747 Chr.19: 49525513-49525513 on Build GRCh38

View Full Product Details