Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156528841-156529114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQGAP3
Entrez Gene ID
128239
Gene Name
IQ motif containing GTPase activating protein 3
Gene Aliases
-
UniGene
Hs.591495
Gene Chromosome Location
Chr.1: 156525405-156572604 on Build GRCh38
Transcripts
NM_178229.4 XM_011509198.2 XM_011509200.2 XM_011509201.2 XM_017000317.1 XM_017000318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
318384 c.4491delG p.T1498fs*80 Chr.1: 156528996-156528996 on Build GRCh38
4405373 c.4442A>C p.K1481T Chr.1: 156529045-156529045 on Build GRCh38
7078548 c.4440G>A p.R1480R Chr.1: 156529047-156529047 on Build GRCh38
6686933 c.4439G>A p.R1480Q Chr.1: 156529048-156529048 on Build GRCh38
325565 c.4490A>T p.K1497M Chr.1: 156528997-156528997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760551838 Chr.1: 156528967-156528967 on Build GRCh38
rs201660294 Chr.1: 156528899-156528899 on Build GRCh38
rs901360228 Chr.1: 156529093-156529093 on Build GRCh38
rs781740971 Chr.1: 156529062-156529062 on Build GRCh38
rs938762819 Chr.1: 156528927-156528927 on Build GRCh38
rs765330384 Chr.1: 156528970-156528970 on Build GRCh38
rs376581680 Chr.1: 156528879-156528879 on Build GRCh38
rs918573301 Chr.1: 156528962-156528962 on Build GRCh38
rs769295448 Chr.1: 156528949-156528949 on Build GRCh38
rs762487661 Chr.1: 156528868-156528868 on Build GRCh38
rs186469606 Chr.1: 156529075-156529075 on Build GRCh38
rs370239156 Chr.1: 156529029-156529029 on Build GRCh38
rs781396792 Chr.1: 156528928-156528928 on Build GRCh38
rs745422432 Chr.1: 156529082-156529082 on Build GRCh38
rs375639213 Chr.1: 156528985-156528985 on Build GRCh38
rs200194524 Chr.1: 156528898-156528898 on Build GRCh38
rs575326348 Chr.1: 156528863-156528863 on Build GRCh38
rs550840918 Chr.1: 156529017-156529017 on Build GRCh38
rs750431254 Chr.1: 156528911-156528911 on Build GRCh38
rs369154566 Chr.1: 156528990-156528990 on Build GRCh38
rs762945955 Chr.1: 156528952-156528952 on Build GRCh38
rs771864676 Chr.1: 156529086-156529086 on Build GRCh38
rs748590937 Chr.1: 156528935-156528935 on Build GRCh38
rs374572830 Chr.1: 156528976-156528976 on Build GRCh38
rs927398930 Chr.1: 156528940-156528940 on Build GRCh38
rs759460179 Chr.1: 156528957-156528957 on Build GRCh38
rs139951838 Chr.1: 156528984-156528984 on Build GRCh38
rs200817577 Chr.1: 156528964-156528964 on Build GRCh38
rs956492665 Chr.1: 156528867-156528867 on Build GRCh38
rs182191022 Chr.1: 156528923-156528923 on Build GRCh38
rs548733563 Chr.1: 156529080-156529080 on Build GRCh38
rs767216073 Chr.1: 156528883-156528883 on Build GRCh38
rs773920999 Chr.1: 156528953-156528953 on Build GRCh38
rs748938834 Chr.1: 156529076-156529076 on Build GRCh38
rs370886023 Chr.1: 156528900-156528900 on Build GRCh38
rs377490024 Chr.1: 156528882-156528882 on Build GRCh38
rs201066805 Chr.1: 156528924-156528924 on Build GRCh38
rs201296692 Chr.1: 156529063-156529063 on Build GRCh38
rs767540369 Chr.1: 156528963-156528963 on Build GRCh38
rs772944929 Chr.1: 156528950-156528950 on Build GRCh38
rs775521236 Chr.1: 156529088-156529088 on Build GRCh38
rs968902257 Chr.1: 156528896-156528896 on Build GRCh38
rs1004503689 Chr.1: 156529059-156529059 on Build GRCh38
rs755414222 Chr.1: 156529048-156529048 on Build GRCh38
rs758575418 Chr.1: 156528920-156528920 on Build GRCh38
rs1171564 Chr.1: 156528881-156528881 on Build GRCh38

View Full Product Details