Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46784183-46784434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESD
Entrez Gene ID
2098
Gene Name
esterase D
Gene Aliases
FGH
UniGene
Hs.432491
Gene Chromosome Location
Chr.13: 46771256-46797681 on Build GRCh38
Transcripts
NM_001984.1 XM_005266278.2 XM_011534954.1 XM_017020414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4681693 c.183delT p.F61fs*72 Chr.13: 46784325-46784325 on Build GRCh38
1942150 c.186A>G p.I62M Chr.13: 46784322-46784322 on Build GRCh38
4474887 c.194C>T p.S65F Chr.13: 46784314-46784314 on Build GRCh38
1639362 c.219A>G p.E73E Chr.13: 46784289-46784289 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781310311 Chr.13: 46784358-46784358 on Build GRCh38
rs749567939 Chr.13: 46784402-46784402 on Build GRCh38
rs146959140 Chr.13: 46784373-46784373 on Build GRCh38
rs766423840 Chr.13: 46784284-46784284 on Build GRCh38
rs757307974 Chr.13: 46784356-46784356 on Build GRCh38
rs200685056 Chr.13: 46784252-46784252 on Build GRCh38
rs148120626 Chr.13: 46784270-46784270 on Build GRCh38
rs770132977 Chr.13: 46784234-46784234 on Build GRCh38
rs748517744 Chr.13: 46784254-46784254 on Build GRCh38
rs546188314 Chr.13: 46784245-46784245 on Build GRCh38
rs370259594 Chr.13: 46784274-46784274 on Build GRCh38
rs774116723 Chr.13: 46784302-46784304 on Build GRCh38
rs777815888 Chr.13: 46784300-46784300 on Build GRCh38
rs943899803 Chr.13: 46784339-46784339 on Build GRCh38
rs575674032 Chr.13: 46784228-46784228 on Build GRCh38
rs754819864 Chr.13: 46784292-46784292 on Build GRCh38
rs11553694 Chr.13: 46784328-46784328 on Build GRCh38
rs746426710 Chr.13: 46784232-46784232 on Build GRCh38
rs141902299 Chr.13: 46784324-46784324 on Build GRCh38
rs781307205 Chr.13: 46784269-46784269 on Build GRCh38
rs763596145 Chr.13: 46784246-46784246 on Build GRCh38
rs767178200 Chr.13: 46784255-46784255 on Build GRCh38
rs746373695 Chr.13: 46784368-46784368 on Build GRCh38
rs147452850 Chr.13: 46784267-46784267 on Build GRCh38
rs775903164 Chr.13: 46784244-46784244 on Build GRCh38
rs8192889 Chr.13: 46784405-46784405 on Build GRCh38
rs774076562 Chr.13: 46784248-46784248 on Build GRCh38
rs770048748 Chr.13: 46784246-46784249 on Build GRCh38
rs753515347 Chr.13: 46784288-46784288 on Build GRCh38
rs897115475 Chr.13: 46784382-46784382 on Build GRCh38
rs775990732 Chr.13: 46784397-46784397 on Build GRCh38

View Full Product Details