Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 73062243-73062500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLF5
Entrez Gene ID
688
Gene Name
Kruppel like factor 5
Gene Aliases
BTEB2, CKLF, IKLF
UniGene
Hs.508234
Gene Chromosome Location
Chr.13: 73054976-73077542 on Build GRCh38
Transcripts
NM_001286818.1 NM_001730.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5435904 c.770C>G p.S257* Chr.13: 73062369-73062369 on Build GRCh38
6554018 c.706G>T p.D236Y Chr.13: 73062305-73062305 on Build GRCh38
5435905 c.834C>G p.F278L Chr.13: 73062433-73062433 on Build GRCh38
5052226 c.822A>T p.A274A Chr.13: 73062421-73062421 on Build GRCh38
3399442 c.864G>A p.M288I Chr.13: 73062463-73062463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746616983 Chr.13: 73062396-73062396 on Build GRCh38
rs765076676 Chr.13: 73062351-73062351 on Build GRCh38
rs772237443 Chr.13: 73062442-73062442 on Build GRCh38
rs35967041 Chr.13: 73062344-73062344 on Build GRCh38
rs374765099 Chr.13: 73062303-73062303 on Build GRCh38
rs770632239 Chr.13: 73062448-73062448 on Build GRCh38
rs372020759 Chr.13: 73062337-73062337 on Build GRCh38
rs756486804 Chr.13: 73062397-73062397 on Build GRCh38
rs139182040 Chr.13: 73062452-73062452 on Build GRCh38
rs1021350397 Chr.13: 73062391-73062391 on Build GRCh38
rs751360624 Chr.13: 73062363-73062374 on Build GRCh38
rs748275920 Chr.13: 73062411-73062411 on Build GRCh38
rs150794367 Chr.13: 73062443-73062443 on Build GRCh38
rs142923337 Chr.13: 73062401-73062401 on Build GRCh38
rs762473554 Chr.13: 73062469-73062469 on Build GRCh38
rs766474046 Chr.13: 73062342-73062342 on Build GRCh38
rs761971376 Chr.13: 73062278-73062278 on Build GRCh38
rs752257469 Chr.13: 73062368-73062368 on Build GRCh38
rs777437255 Chr.13: 73062382-73062382 on Build GRCh38
rs567420958 Chr.13: 73062341-73062341 on Build GRCh38
rs373255152 Chr.13: 73062317-73062317 on Build GRCh38
rs368263038 Chr.13: 73062410-73062410 on Build GRCh38
rs780398922 Chr.13: 73062399-73062399 on Build GRCh38
rs769263806 Chr.13: 73062402-73062402 on Build GRCh38
rs760920273 Chr.13: 73062446-73062446 on Build GRCh38
rs377157891 Chr.13: 73062380-73062380 on Build GRCh38
rs765261626 Chr.13: 73062461-73062461 on Build GRCh38
rs773535185 Chr.13: 73062324-73062324 on Build GRCh38
rs752594595 Chr.13: 73062468-73062468 on Build GRCh38
rs990980354 Chr.13: 73062445-73062445 on Build GRCh38
rs903436534 Chr.13: 73062473-73062473 on Build GRCh38
rs754874654 Chr.13: 73062348-73062348 on Build GRCh38
rs959527660 Chr.13: 73062444-73062444 on Build GRCh38
rs955309434 Chr.13: 73062334-73062334 on Build GRCh38
rs759234494 Chr.13: 73062454-73062454 on Build GRCh38

View Full Product Details