Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134248325-134248596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THYN1
Entrez Gene ID
29087
Gene Name
thymocyte nuclear protein 1
Gene Aliases
HSPC144, MDS012, MY105, THY28, THY28KD
UniGene
Hs.13645
Gene Chromosome Location
Chr.11: 134248278-134253401 on Build GRCh38
Transcripts
NM_001037304.1 NM_001037305.1 NM_014174.2 NM_199297.1 NM_199298.1 XM_005271527.3 XM_005271528.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4647285 c.647T>C p.V216A Chr.11: 134248469-134248469 on Build GRCh38
6583490 c.632-4G>T p.? Chr.11: 134248488-134248488 on Build GRCh38
6560673 c.632-3_632-2insT p.? Chr.11: 134248486-134248487 on Build GRCh38
5942981 c.632-4_632-3insT p.? Chr.11: 134248487-134248488 on Build GRCh38
4019536 c.654C>A p.S218R Chr.11: 134248462-134248462 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs937401400 Chr.11: 134248411-134248411 on Build GRCh38
rs147345168 Chr.11: 134248499-134248499 on Build GRCh38
rs371647723 Chr.11: 134248392-134248392 on Build GRCh38
rs867738661 Chr.11: 134248450-134248450 on Build GRCh38
rs774791264 Chr.11: 134248406-134248406 on Build GRCh38
rs752264081 Chr.11: 134248515-134248516 on Build GRCh38
rs776530634 Chr.11: 134248389-134248389 on Build GRCh38
rs757185816 Chr.11: 134248366-134248366 on Build GRCh38
rs753762617 Chr.11: 134248434-134248434 on Build GRCh38
rs745703087 Chr.11: 134248467-134248467 on Build GRCh38
rs367598913 Chr.11: 134248429-134248429 on Build GRCh38
rs778772392 Chr.11: 134248368-134248368 on Build GRCh38
rs569044762 Chr.11: 134248408-134248408 on Build GRCh38
rs765674963 Chr.11: 134248476-134248477 on Build GRCh38
rs767232016 Chr.11: 134248413-134248413 on Build GRCh38
rs774540320 Chr.11: 134248536-134248536 on Build GRCh38
rs756080653 Chr.11: 134248355-134248355 on Build GRCh38
rs770267462 Chr.11: 134248393-134248393 on Build GRCh38
rs112434812 Chr.11: 134248465-134248465 on Build GRCh38
rs749738565 Chr.11: 134248525-134248525 on Build GRCh38
rs777507397 Chr.11: 134248395-134248396 on Build GRCh38
rs1029861807 Chr.11: 134248573-134248573 on Build GRCh38
rs1041973104 Chr.11: 134248490-134248490 on Build GRCh38
rs773769438 Chr.11: 134248394-134248394 on Build GRCh38
rs377165872 Chr.11: 134248459-134248459 on Build GRCh38
rs763307815 Chr.11: 134248395-134248395 on Build GRCh38
rs769931823 Chr.11: 134248492-134248492 on Build GRCh38
rs371682845 Chr.11: 134248431-134248431 on Build GRCh38
rs773708968 Chr.11: 134248489-134248489 on Build GRCh38
rs755742051 Chr.11: 134248430-134248430 on Build GRCh38
rs531939676 Chr.11: 134248387-134248387 on Build GRCh38
rs553782803 Chr.11: 134248562-134248562 on Build GRCh38
rs1000463529 Chr.11: 134248494-134248494 on Build GRCh38
rs920237900 Chr.11: 134248542-134248542 on Build GRCh38
rs930333593 Chr.11: 134248549-134248549 on Build GRCh38
rs752123827 Chr.11: 134248425-134248425 on Build GRCh38
rs926370507 Chr.11: 134248574-134248574 on Build GRCh38
rs747138876 Chr.11: 134248381-134248381 on Build GRCh38
rs571961308 Chr.11: 134248491-134248491 on Build GRCh38
rs911789051 Chr.11: 134248496-134248496 on Build GRCh38
rs771492807 Chr.11: 134248531-134248531 on Build GRCh38
rs550169661 Chr.11: 134248480-134248480 on Build GRCh38
rs752275569 Chr.11: 134248353-134248353 on Build GRCh38
rs556541008 Chr.11: 134248356-134248356 on Build GRCh38
rs71479599 Chr.11: 134248488-134248489 on Build GRCh38
rs539387596 Chr.11: 134248511-134248511 on Build GRCh38
rs766589603 Chr.11: 134248396-134248396 on Build GRCh38
rs757269932 Chr.11: 134248456-134248456 on Build GRCh38
rs953205881 Chr.11: 134248481-134248481 on Build GRCh38
rs749079973 Chr.11: 134248365-134248365 on Build GRCh38
rs376209516 Chr.11: 134248422-134248422 on Build GRCh38

View Full Product Details