Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76680742-76681003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MXRA7
Entrez Gene ID
439921
Gene Name
matrix remodeling associated 7
Gene Aliases
PS1TP1, TMAP1
UniGene
Hs.250723
Gene Chromosome Location
Chr.17: 76673650-76711005 on Build GRCh38
Transcripts
NM_001008528.2 NM_001008529.2 NM_198530.3 NR_130926.1 NR_130927.1 NR_130928.1 XM_005257382.2 XM_005257383.2 XM_005257384.2 XM_011524830.1

SNP Detail

SNP ID Chromosome Location
rs377011607 Chr.17: 76680895-76680895 on Build GRCh38
rs751916433 Chr.17: 76680889-76680889 on Build GRCh38
rs756895808 Chr.17: 76680887-76680887 on Build GRCh38
rs948878885 Chr.17: 76680905-76680905 on Build GRCh38
rs932556180 Chr.17: 76680955-76680955 on Build GRCh38
rs1028231041 Chr.17: 76680804-76680804 on Build GRCh38
rs922273456 Chr.17: 76680946-76680946 on Build GRCh38
rs907461289 Chr.17: 76680977-76680977 on Build GRCh38
rs375675033 Chr.17: 76680837-76680837 on Build GRCh38
rs781719062 Chr.17: 76680904-76680904 on Build GRCh38
rs747411198 Chr.17: 76680825-76680825 on Build GRCh38
rs561766378 Chr.17: 76680865-76680865 on Build GRCh38
rs767361610 Chr.17: 76680866-76680866 on Build GRCh38
rs552665666 Chr.17: 76680766-76680766 on Build GRCh38
rs760571611 Chr.17: 76680869-76680869 on Build GRCh38
rs755388582 Chr.17: 76680899-76680899 on Build GRCh38
rs753311362 Chr.17: 76680883-76680883 on Build GRCh38
rs200192186 Chr.17: 76680890-76680890 on Build GRCh38
rs368899347 Chr.17: 76680850-76680850 on Build GRCh38
rs769220001 Chr.17: 76680828-76680828 on Build GRCh38
rs748664615 Chr.17: 76680906-76680906 on Build GRCh38
rs770223430 Chr.17: 76680919-76680919 on Build GRCh38
rs769884318 Chr.17: 76680842-76680842 on Build GRCh38
rs771087675 Chr.17: 76680853-76680853 on Build GRCh38
rs781263063 Chr.17: 76680790-76680790 on Build GRCh38
rs530720017 Chr.17: 76680870-76680870 on Build GRCh38
rs777865932 Chr.17: 76680927-76680927 on Build GRCh38
rs146537764 Chr.17: 76680965-76680965 on Build GRCh38
rs952275202 Chr.17: 76680836-76680836 on Build GRCh38
rs372980214 Chr.17: 76680851-76680851 on Build GRCh38
rs753897657 Chr.17: 76680900-76680900 on Build GRCh38
rs774061429 Chr.17: 76680855-76680855 on Build GRCh38
rs748731471 Chr.17: 76680839-76680839 on Build GRCh38
rs572812419 Chr.17: 76680774-76680774 on Build GRCh38
rs976352978 Chr.17: 76680923-76680923 on Build GRCh38
rs545761230 Chr.17: 76680762-76680762 on Build GRCh38

View Full Product Details