Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 17284570-17284811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM24
Entrez Gene ID
221662
Gene Name
RNA binding motif protein 24
Gene Aliases
RNPC6, dJ259A10.1
UniGene
Hs.519904
Gene Chromosome Location
Chr.6: 17281508-17293875 on Build GRCh38
Transcripts
NM_001143941.1 NM_001143942.1 NM_153020.2 XM_005248932.3 XM_011514387.2 XM_011514388.2 XM_011514389.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3623027 c.319C>T p.H107Y Chr.6: 17284683-17284683 on Build GRCh38
4750487 c.211G>A p.G71R Chr.6: 17284710-17284710 on Build GRCh38
76190 c.212G>A p.G71E Chr.6: 17284711-17284711 on Build GRCh38
6247201 c.293-5G>T p.? Chr.6: 17284652-17284652 on Build GRCh38
3430355 c.345C>T p.F115F Chr.6: 17284709-17284709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542695169 Chr.6: 17284751-17284751 on Build GRCh38
rs779628629 Chr.6: 17284730-17284730 on Build GRCh38
rs754507184 Chr.6: 17284746-17284746 on Build GRCh38
rs765441614 Chr.6: 17284689-17284689 on Build GRCh38
rs267600899 Chr.6: 17284705-17284705 on Build GRCh38
rs200835058 Chr.6: 17284710-17284710 on Build GRCh38
rs538560522 Chr.6: 17284735-17284739 on Build GRCh38
rs766514290 Chr.6: 17284655-17284655 on Build GRCh38
rs773285641 Chr.6: 17284758-17284759 on Build GRCh38
rs774201472 Chr.6: 17284610-17284610 on Build GRCh38
rs771896921 Chr.6: 17284750-17284750 on Build GRCh38
rs989220631 Chr.6: 17284650-17284650 on Build GRCh38
rs747917150 Chr.6: 17284630-17284633 on Build GRCh38
rs374717714 Chr.6: 17284696-17284696 on Build GRCh38
rs539148835 Chr.6: 17284641-17284641 on Build GRCh38
rs773184470 Chr.6: 17284637-17284637 on Build GRCh38
rs574698991 Chr.6: 17284765-17284765 on Build GRCh38
rs949125413 Chr.6: 17284663-17284663 on Build GRCh38
rs371735746 Chr.6: 17284729-17284729 on Build GRCh38
rs771843450 Chr.6: 17284635-17284635 on Build GRCh38
rs200637339 Chr.6: 17284715-17284715 on Build GRCh38
rs185780622 Chr.6: 17284747-17284747 on Build GRCh38
rs138850194 Chr.6: 17284709-17284709 on Build GRCh38
rs748675531 Chr.6: 17284779-17284779 on Build GRCh38
rs760578074 Chr.6: 17284639-17284639 on Build GRCh38
rs143113042 Chr.6: 17284683-17284683 on Build GRCh38
rs748004665 Chr.6: 17284616-17284616 on Build GRCh38
rs753221169 Chr.6: 17284742-17284742 on Build GRCh38
rs776528557 Chr.6: 17284679-17284679 on Build GRCh38
rs752141415 Chr.6: 17284723-17284723 on Build GRCh38
rs907849025 Chr.6: 17284769-17284769 on Build GRCh38
rs747777628 Chr.6: 17284749-17284749 on Build GRCh38

View Full Product Details