Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165295885-165296120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN2A
Entrez Gene ID
6326
Gene Name
sodium voltage-gated channel alpha subunit 2
Gene Aliases
BFIC3, BFIS3, BFNIS, EIEE11, HBA, HBSCI, HBSCII, NAC2, Na(v)1.2, Nav1.2, SCN2A1, SCN2A2
UniGene
Hs.93485
Gene Chromosome Location
Chr.2: 165208056-165392310 on Build GRCh38
Transcripts
NM_001040142.1 NM_001040143.1 NM_021007.2 XM_005246753.3 XM_017004655.1 XM_017004656.1 XM_017004657.1 XM_017004658.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5736237 c.64C>A p.L22I Chr.2: 165295887-165295887 on Build GRCh38
3312288 c.264G>T p.K88N Chr.2: 165296087-165296087 on Build GRCh38
4305090 c.132G>A p.E44E Chr.2: 165295955-165295955 on Build GRCh38
1008552 c.105G>T p.K35N Chr.2: 165295928-165295928 on Build GRCh38
3312289 c.264G>T p.K88N Chr.2: 165296087-165296087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754993031 Chr.2: 165295945-165295945 on Build GRCh38
rs557687080 Chr.2: 165295983-165295983 on Build GRCh38
rs536682525 Chr.2: 165296085-165296085 on Build GRCh38
rs773605666 Chr.2: 165295968-165295968 on Build GRCh38
rs757194863 Chr.2: 165295938-165295938 on Build GRCh38
rs781272663 Chr.2: 165295955-165295955 on Build GRCh38
rs749676317 Chr.2: 165295977-165295977 on Build GRCh38
rs144814658 Chr.2: 165295923-165295923 on Build GRCh38
rs778773546 Chr.2: 165295939-165295939 on Build GRCh38
rs771468513 Chr.2: 165296039-165296039 on Build GRCh38
rs1007052146 Chr.2: 165295906-165295906 on Build GRCh38
rs796053110 Chr.2: 165296000-165296000 on Build GRCh38
rs748331725 Chr.2: 165295957-165295957 on Build GRCh38
rs763906285 Chr.2: 165295932-165295932 on Build GRCh38
rs962766268 Chr.2: 165295959-165295959 on Build GRCh38
rs143065769 Chr.2: 165296075-165296075 on Build GRCh38
rs34411329 Chr.2: 165296055-165296055 on Build GRCh38
rs796053168 Chr.2: 165296043-165296043 on Build GRCh38
rs370552463 Chr.2: 165295909-165295909 on Build GRCh38
rs796053167 Chr.2: 165295929-165295929 on Build GRCh38
rs145233994 Chr.2: 165295961-165295961 on Build GRCh38
rs761753124 Chr.2: 165295933-165295934 on Build GRCh38
rs200479065 Chr.2: 165296060-165296060 on Build GRCh38
rs747086776 Chr.2: 165295944-165295944 on Build GRCh38
rs199657941 Chr.2: 165296078-165296078 on Build GRCh38
rs753614123 Chr.2: 165295937-165295937 on Build GRCh38

View Full Product Details