Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 141790830-141791077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAS2R5
Entrez Gene ID
54429
Gene Name
taste 2 receptor member 5
Gene Aliases
T2R5
UniGene
Hs.675370
Gene Chromosome Location
Chr.7: 141790217-141791366 on Build GRCh38
Transcripts
NM_018980.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3634324 c.612C>T p.V204V Chr.7: 141790973-141790973 on Build GRCh38
6847662 c.597C>A p.H199Q Chr.7: 141790958-141790958 on Build GRCh38
1699460 c.637C>T p.R213W Chr.7: 141790998-141790998 on Build GRCh38
5675820 c.705C>G p.I235M Chr.7: 141791066-141791066 on Build GRCh38
5644435 c.629G>A p.R210K Chr.7: 141790990-141790990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775420198 Chr.7: 141790914-141790914 on Build GRCh38
rs761343280 Chr.7: 141790953-141790953 on Build GRCh38
rs760653413 Chr.7: 141790921-141790921 on Build GRCh38
rs2234015 Chr.7: 141790999-141790999 on Build GRCh38
rs773893203 Chr.7: 141790932-141790932 on Build GRCh38
rs147451663 Chr.7: 141790882-141790882 on Build GRCh38
rs149987830 Chr.7: 141790998-141790998 on Build GRCh38
rs144096933 Chr.7: 141791041-141791041 on Build GRCh38
rs747180926 Chr.7: 141791043-141791043 on Build GRCh38
rs986010781 Chr.7: 141790872-141790872 on Build GRCh38
rs4726476 Chr.7: 141790987-141790987 on Build GRCh38
rs753917011 Chr.7: 141790943-141790944 on Build GRCh38
rs34529840 Chr.7: 141790861-141790861 on Build GRCh38
rs377498979 Chr.7: 141790859-141790859 on Build GRCh38
rs779112677 Chr.7: 141790855-141790855 on Build GRCh38
rs766509244 Chr.7: 141790928-141790933 on Build GRCh38
rs755350481 Chr.7: 141791015-141791015 on Build GRCh38
rs1028296845 Chr.7: 141790899-141790899 on Build GRCh38
rs957439122 Chr.7: 141790906-141790906 on Build GRCh38
rs534109459 Chr.7: 141790957-141790957 on Build GRCh38
rs559569114 Chr.7: 141790886-141790886 on Build GRCh38
rs543303670 Chr.7: 141790917-141790917 on Build GRCh38
rs988807473 Chr.7: 141790938-141790938 on Build GRCh38
rs139239350 Chr.7: 141790989-141790989 on Build GRCh38
rs758688445 Chr.7: 141791023-141791023 on Build GRCh38
rs745590916 Chr.7: 141790978-141790978 on Build GRCh38
rs762817573 Chr.7: 141790986-141790988 on Build GRCh38
rs761160402 Chr.7: 141790992-141790992 on Build GRCh38
rs768362101 Chr.7: 141790924-141790924 on Build GRCh38
rs371414884 Chr.7: 141790874-141790874 on Build GRCh38
rs756048842 Chr.7: 141791053-141791053 on Build GRCh38
rs148483148 Chr.7: 141791021-141791021 on Build GRCh38
rs765450946 Chr.7: 141791020-141791020 on Build GRCh38
rs866855261 Chr.7: 141790921-141790921 on Build GRCh38

View Full Product Details