Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212342145-212342406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP2R5A
Entrez Gene ID
5525
Gene Name
protein phosphatase 2 regulatory subunit B'alpha
Gene Aliases
B56A, PR61A
UniGene
Hs.744012
Gene Chromosome Location
Chr.1: 212285537-212361863 on Build GRCh38
Transcripts
NM_001199756.1 NM_006243.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4391496 c.490G>T p.E164* Chr.1: 212342197-212342197 on Build GRCh38
6211939 c.377A>G p.Y126C Chr.1: 212342255-212342255 on Build GRCh38
5478054 c.495C>A p.F165L Chr.1: 212342202-212342202 on Build GRCh38
5478055 c.324C>A p.F108L Chr.1: 212342202-212342202 on Build GRCh38
4391497 c.319G>T p.E107* Chr.1: 212342197-212342197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776597036 Chr.1: 212342352-212342352 on Build GRCh38
rs778096953 Chr.1: 212342199-212342199 on Build GRCh38
rs747560993 Chr.1: 212342222-212342222 on Build GRCh38
rs776766301 Chr.1: 212342243-212342243 on Build GRCh38
rs775228993 Chr.1: 212342206-212342206 on Build GRCh38
rs184824973 Chr.1: 212342291-212342291 on Build GRCh38
rs779904784 Chr.1: 212342181-212342181 on Build GRCh38
rs772598006 Chr.1: 212342188-212342188 on Build GRCh38
rs931751731 Chr.1: 212342278-212342278 on Build GRCh38
rs370623113 Chr.1: 212342328-212342328 on Build GRCh38
rs761463568 Chr.1: 212342292-212342293 on Build GRCh38
rs778200025 Chr.1: 212342326-212342326 on Build GRCh38
rs1019317206 Chr.1: 212342241-212342241 on Build GRCh38
rs551205285 Chr.1: 212342302-212342302 on Build GRCh38
rs755727845 Chr.1: 212342306-212342306 on Build GRCh38
rs1774249 Chr.1: 212342271-212342271 on Build GRCh38
rs1046603153 Chr.1: 212342339-212342341 on Build GRCh38
rs374082551 Chr.1: 212342260-212342260 on Build GRCh38
rs770844766 Chr.1: 212342223-212342223 on Build GRCh38
rs757765765 Chr.1: 212342330-212342330 on Build GRCh38
rs749927902 Chr.1: 212342301-212342301 on Build GRCh38
rs941673642 Chr.1: 212342319-212342319 on Build GRCh38
rs762696931 Chr.1: 212342268-212342268 on Build GRCh38
rs17852039 Chr.1: 212342234-212342234 on Build GRCh38
rs34910845 Chr.1: 212342216-212342216 on Build GRCh38
rs965269011 Chr.1: 212342255-212342255 on Build GRCh38
rs147962315 Chr.1: 212342272-212342272 on Build GRCh38
rs377603944 Chr.1: 212342325-212342325 on Build GRCh38
rs192765953 Chr.1: 212342252-212342252 on Build GRCh38
rs753571645 Chr.1: 212342323-212342323 on Build GRCh38
rs766091789 Chr.1: 212342310-212342310 on Build GRCh38
rs369698862 Chr.1: 212342257-212342257 on Build GRCh38

View Full Product Details