Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100719772-100720023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VCAM1
Entrez Gene ID
7412
Gene Name
vascular cell adhesion molecule 1
Gene Aliases
CD106, INCAM-100
UniGene
Hs.109225
Gene Chromosome Location
Chr.1: 100719640-100739045 on Build GRCh38
Transcripts
NM_001078.3 NM_001199834.1 NM_080682.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6879792 c.24C>T p.I8I Chr.1: 100719884-100719884 on Build GRCh38
5988837 c.59C>G p.A20G Chr.1: 100719919-100719919 on Build GRCh38
5015884 c.9_10insA p.M5fs*22 Chr.1: 100719869-100719870 on Build GRCh38
3470312 c.9G>A p.G3G Chr.1: 100719869-100719869 on Build GRCh38
6255963 c.47G>T p.W16L Chr.1: 100719907-100719907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932949729 Chr.1: 100719798-100719798 on Build GRCh38
rs754145648 Chr.1: 100719887-100719887 on Build GRCh38
rs1050695808 Chr.1: 100719881-100719881 on Build GRCh38
rs375980259 Chr.1: 100719915-100719915 on Build GRCh38
rs779494308 Chr.1: 100719851-100719851 on Build GRCh38
rs752103980 Chr.1: 100719932-100719932 on Build GRCh38
rs757653671 Chr.1: 100719837-100719837 on Build GRCh38
rs201966059 Chr.1: 100719834-100719834 on Build GRCh38
rs140406427 Chr.1: 100719938-100719938 on Build GRCh38
rs3783610 Chr.1: 100719847-100719847 on Build GRCh38
rs34228330 Chr.1: 100719914-100719914 on Build GRCh38
rs764674964 Chr.1: 100719832-100719832 on Build GRCh38
rs144133931 Chr.1: 100719897-100719897 on Build GRCh38
rs767930526 Chr.1: 100719941-100719941 on Build GRCh38
rs1054626081 Chr.1: 100719843-100719843 on Build GRCh38
rs774366600 Chr.1: 100719855-100719855 on Build GRCh38
rs202092399 Chr.1: 100719900-100719900 on Build GRCh38
rs748694342 Chr.1: 100719974-100719974 on Build GRCh38
rs762922546 Chr.1: 100719797-100719797 on Build GRCh38
rs750067959 Chr.1: 100719942-100719942 on Build GRCh38
rs755693248 Chr.1: 100719950-100719950 on Build GRCh38
rs201513859 Chr.1: 100719854-100719854 on Build GRCh38
rs1002803049 Chr.1: 100719985-100719987 on Build GRCh38
rs148280495 Chr.1: 100719891-100719891 on Build GRCh38
rs747911510 Chr.1: 100719877-100719877 on Build GRCh38
rs780793820 Chr.1: 100719841-100719841 on Build GRCh38
rs546776871 Chr.1: 100719965-100719965 on Build GRCh38
rs775374597 Chr.1: 100719903-100719903 on Build GRCh38
rs3783609 Chr.1: 100719807-100719807 on Build GRCh38
rs368721099 Chr.1: 100719820-100719820 on Build GRCh38
rs533068429 Chr.1: 100719951-100719951 on Build GRCh38
rs903241400 Chr.1: 100719981-100719981 on Build GRCh38
rs184836168 Chr.1: 100719922-100719922 on Build GRCh38
rs772031058 Chr.1: 100719883-100719883 on Build GRCh38

View Full Product Details