Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154323562-154323831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP10
Entrez Gene ID
89872
Gene Name
aquaporin 10
Gene Aliases
AQPA_HUMAN
UniGene
Hs.259048
Gene Chromosome Location
Chr.1: 154321059-154325325 on Build GRCh38
Transcripts
NM_080429.2 XM_011510104.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4924906 c.534G>C p.L178L Chr.1: 154323633-154323633 on Build GRCh38
3474662 c.611C>T p.S204F Chr.1: 154323710-154323710 on Build GRCh38
6888386 c.642C>T p.N214N Chr.1: 154323741-154323741 on Build GRCh38
3705022 c.541C>T p.R181W Chr.1: 154323640-154323640 on Build GRCh38
1688668 c.584G>A p.G195E Chr.1: 154323683-154323683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781413989 Chr.1: 154323611-154323611 on Build GRCh38
rs751048077 Chr.1: 154323702-154323702 on Build GRCh38
rs747692921 Chr.1: 154323661-154323661 on Build GRCh38
rs867427480 Chr.1: 154323741-154323741 on Build GRCh38
rs751948567 Chr.1: 154323602-154323602 on Build GRCh38
rs767755990 Chr.1: 154323738-154323738 on Build GRCh38
rs746786961 Chr.1: 154323777-154323777 on Build GRCh38
rs745857248 Chr.1: 154323641-154323641 on Build GRCh38
rs200639079 Chr.1: 154323764-154323764 on Build GRCh38
rs376171472 Chr.1: 154323654-154323654 on Build GRCh38
rs949215887 Chr.1: 154323655-154323655 on Build GRCh38
rs752776491 Chr.1: 154323616-154323616 on Build GRCh38
rs898792414 Chr.1: 154323773-154323773 on Build GRCh38
rs193133403 Chr.1: 154323746-154323746 on Build GRCh38
rs768207751 Chr.1: 154323659-154323659 on Build GRCh38
rs758960762 Chr.1: 154323595-154323595 on Build GRCh38
rs377590602 Chr.1: 154323778-154323778 on Build GRCh38
rs778846975 Chr.1: 154323640-154323640 on Build GRCh38
rs934672125 Chr.1: 154323589-154323589 on Build GRCh38
rs372825269 Chr.1: 154323760-154323760 on Build GRCh38
rs759873653 Chr.1: 154323731-154323731 on Build GRCh38
rs758326907 Chr.1: 154323766-154323766 on Build GRCh38
rs750910346 Chr.1: 154323588-154323588 on Build GRCh38
rs752864581 Chr.1: 154323742-154323742 on Build GRCh38
rs370728661 Chr.1: 154323800-154323800 on Build GRCh38
rs144627448 Chr.1: 154323804-154323804 on Build GRCh38
rs761432606 Chr.1: 154323659-154323660 on Build GRCh38
rs756143604 Chr.1: 154323631-154323631 on Build GRCh38
rs370543262 Chr.1: 154323660-154323660 on Build GRCh38
rs755209873 Chr.1: 154323604-154323604 on Build GRCh38
rs779984854 Chr.1: 154323768-154323768 on Build GRCh38
rs779783182 Chr.1: 154323652-154323652 on Build GRCh38
rs147224722 Chr.1: 154323630-154323630 on Build GRCh38
rs772816368 Chr.1: 154323697-154323697 on Build GRCh38
rs774589159 Chr.1: 154323727-154323727 on Build GRCh38
rs772020135 Chr.1: 154323648-154323648 on Build GRCh38
rs753952871 Chr.1: 154323763-154323763 on Build GRCh38
rs1194610 Chr.1: 154323600-154323600 on Build GRCh38
rs769477393 Chr.1: 154323675-154323675 on Build GRCh38
rs559006639 Chr.1: 154323726-154323726 on Build GRCh38
rs1026061558 Chr.1: 154323806-154323806 on Build GRCh38
rs756309400 Chr.1: 154323749-154323749 on Build GRCh38
rs765233123 Chr.1: 154323761-154323762 on Build GRCh38

View Full Product Details