Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49968558-49968819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB113
Entrez Gene ID
245927
Gene Name
defensin beta 113
Gene Aliases
DEFB-13
UniGene
Hs.582789
Gene Chromosome Location
Chr.6: 49968677-49969625 on Build GRCh38
Transcripts
NM_001037729.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
596728 c.147G>A p.W49* Chr.6: 49968779-49968779 on Build GRCh38
1080107 c.143G>A p.S48N Chr.6: 49968783-49968783 on Build GRCh38
4356290 c.170A>G p.N57S Chr.6: 49968756-49968756 on Build GRCh38
1445025 c.132G>A p.P44P Chr.6: 49968794-49968794 on Build GRCh38
1445026 c.116G>A p.R39H Chr.6: 49968810-49968810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752895772 Chr.6: 49968794-49968794 on Build GRCh38
rs200286174 Chr.6: 49968780-49968780 on Build GRCh38
rs373909445 Chr.6: 49968746-49968746 on Build GRCh38
rs376862450 Chr.6: 49968749-49968749 on Build GRCh38
rs570816422 Chr.6: 49968610-49968610 on Build GRCh38
rs779312079 Chr.6: 49968737-49968737 on Build GRCh38
rs751454097 Chr.6: 49968776-49968776 on Build GRCh38
rs372608145 Chr.6: 49968724-49968724 on Build GRCh38
rs770546470 Chr.6: 49968643-49968643 on Build GRCh38
rs781350427 Chr.6: 49968701-49968701 on Build GRCh38
rs749714531 Chr.6: 49968727-49968727 on Build GRCh38
rs748737381 Chr.6: 49968622-49968622 on Build GRCh38
rs377587500 Chr.6: 49968796-49968796 on Build GRCh38
rs755042267 Chr.6: 49968699-49968699 on Build GRCh38
rs759456897 Chr.6: 49968779-49968779 on Build GRCh38
rs958464994 Chr.6: 49968790-49968790 on Build GRCh38
rs548800329 Chr.6: 49968615-49968618 on Build GRCh38
rs201896487 Chr.6: 49968800-49968800 on Build GRCh38
rs369144899 Chr.6: 49968713-49968713 on Build GRCh38
rs371006158 Chr.6: 49968788-49968788 on Build GRCh38
rs555913466 Chr.6: 49968760-49968760 on Build GRCh38
rs769476112 Chr.6: 49968754-49968754 on Build GRCh38
rs762677670 Chr.6: 49968757-49968757 on Build GRCh38
rs879150748 Chr.6: 49968709-49968709 on Build GRCh38
rs765031187 Chr.6: 49968642-49968642 on Build GRCh38
rs751463780 Chr.6: 49968696-49968696 on Build GRCh38
rs201564508 Chr.6: 49968738-49968738 on Build GRCh38
rs201224982 Chr.6: 49968795-49968795 on Build GRCh38
rs1007625845 Chr.6: 49968690-49968690 on Build GRCh38
rs752761143 Chr.6: 49968633-49968634 on Build GRCh38
rs773844292 Chr.6: 49968666-49968666 on Build GRCh38
rs200246441 Chr.6: 49968732-49968732 on Build GRCh38
rs776062433 Chr.6: 49968748-49968748 on Build GRCh38
rs79563352 Chr.6: 49968651-49968651 on Build GRCh38
rs928554760 Chr.6: 49968609-49968609 on Build GRCh38
rs180815205 Chr.6: 49968756-49968756 on Build GRCh38
rs778141088 Chr.6: 49968726-49968726 on Build GRCh38
rs767544216 Chr.6: 49968784-49968784 on Build GRCh38
rs188690075 Chr.6: 49968618-49968618 on Build GRCh38

View Full Product Details