Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120905033-120905264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAF1
Entrez Gene ID
7185
Gene Name
TNF receptor associated factor 1
Gene Aliases
EBI6, MGC:10353
UniGene
Hs.531251
Gene Chromosome Location
Chr.9: 120902393-120929173 on Build GRCh38
Transcripts
NM_001190945.1 NM_001190947.1 NM_005658.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5586842 c.1100G>A p.S367N Chr.9: 120905171-120905171 on Build GRCh38
1104698 c.1046T>C p.L349P Chr.9: 120905225-120905225 on Build GRCh38
146038 c.1136A>G p.N379S Chr.9: 120905135-120905135 on Build GRCh38
6341226 c.1081G>T p.A361S Chr.9: 120905190-120905190 on Build GRCh38
1569441 c.1201G>A p.V401M Chr.9: 120905070-120905070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377159992 Chr.9: 120905200-120905200 on Build GRCh38
rs751835432 Chr.9: 120905086-120905086 on Build GRCh38
rs935043914 Chr.9: 120905176-120905176 on Build GRCh38
rs371039591 Chr.9: 120905070-120905070 on Build GRCh38
rs767693183 Chr.9: 120905096-120905096 on Build GRCh38
rs138123440 Chr.9: 120905062-120905062 on Build GRCh38
rs745699808 Chr.9: 120905214-120905214 on Build GRCh38
rs199835146 Chr.9: 120905184-120905184 on Build GRCh38
rs752456981 Chr.9: 120905137-120905137 on Build GRCh38
rs759834952 Chr.9: 120905190-120905190 on Build GRCh38
rs746485879 Chr.9: 120905239-120905239 on Build GRCh38
rs758021473 Chr.9: 120905218-120905218 on Build GRCh38
rs149591033 Chr.9: 120905065-120905065 on Build GRCh38
rs767894014 Chr.9: 120905191-120905191 on Build GRCh38
rs148362756 Chr.9: 120905077-120905077 on Build GRCh38
rs775035728 Chr.9: 120905064-120905064 on Build GRCh38
rs992352655 Chr.9: 120905090-120905090 on Build GRCh38
rs768054582 Chr.9: 120905241-120905241 on Build GRCh38
rs752893475 Chr.9: 120905115-120905115 on Build GRCh38
rs983526021 Chr.9: 120905195-120905195 on Build GRCh38
rs750776538 Chr.9: 120905071-120905071 on Build GRCh38
rs772658437 Chr.9: 120905134-120905134 on Build GRCh38
rs771774293 Chr.9: 120905145-120905145 on Build GRCh38
rs766666979 Chr.9: 120905183-120905183 on Build GRCh38
rs916860915 Chr.9: 120905103-120905103 on Build GRCh38
rs774946070 Chr.9: 120905161-120905161 on Build GRCh38
rs267602110 Chr.9: 120905110-120905110 on Build GRCh38
rs376450570 Chr.9: 120905233-120905233 on Build GRCh38
rs147544712 Chr.9: 120905181-120905181 on Build GRCh38
rs770284947 Chr.9: 120905165-120905165 on Build GRCh38
rs752805562 Chr.9: 120905199-120905199 on Build GRCh38
rs755051203 Chr.9: 120905094-120905094 on Build GRCh38
rs377746528 Chr.9: 120905207-120905207 on Build GRCh38
rs368165927 Chr.9: 120905164-120905164 on Build GRCh38
rs777622191 Chr.9: 120905119-120905119 on Build GRCh38
rs143200014 Chr.9: 120905133-120905133 on Build GRCh38
rs1039254922 Chr.9: 120905210-120905210 on Build GRCh38
rs143265058 Chr.9: 120905076-120905076 on Build GRCh38
rs201773418 Chr.9: 120905208-120905208 on Build GRCh38
rs371622560 Chr.9: 120905135-120905135 on Build GRCh38
rs756049894 Chr.9: 120905116-120905116 on Build GRCh38

View Full Product Details