Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28547843-28548104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNC119
Entrez Gene ID
9094
Gene Name
unc-119 lipid binding chaperone
Gene Aliases
HRG4, IMD13, POC7, POC7A
UniGene
Hs.410455
Gene Chromosome Location
Chr.17: 28546707-28552633 on Build GRCh38
Transcripts
NM_005148.3 NM_054035.2 XM_011525459.2 XM_017025289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
242172 c.416G>A p.R139H Chr.17: 28548020-28548020 on Build GRCh38
4751569 c.355C>T p.R119W Chr.17: 28548081-28548081 on Build GRCh38
5023784 c.340T>G p.L114V Chr.17: 28548096-28548096 on Build GRCh38
4739215 c.356G>A p.R119Q Chr.17: 28548080-28548080 on Build GRCh38
4739217 c.337C>T p.R113W Chr.17: 28548099-28548099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762825360 Chr.17: 28547874-28547874 on Build GRCh38
rs757339891 Chr.17: 28547881-28547881 on Build GRCh38
rs770534020 Chr.17: 28547875-28547876 on Build GRCh38
rs201337554 Chr.17: 28548020-28548020 on Build GRCh38
rs905472030 Chr.17: 28547928-28547928 on Build GRCh38
rs535665290 Chr.17: 28548009-28548009 on Build GRCh38
rs141782779 Chr.17: 28548032-28548032 on Build GRCh38
rs200895223 Chr.17: 28548048-28548048 on Build GRCh38
rs776489945 Chr.17: 28547980-28547980 on Build GRCh38
rs778180208 Chr.17: 28547975-28547975 on Build GRCh38
rs1046078658 Chr.17: 28547922-28547922 on Build GRCh38
rs370221268 Chr.17: 28547991-28547991 on Build GRCh38
rs376844875 Chr.17: 28547872-28547872 on Build GRCh38
rs769842915 Chr.17: 28548060-28548060 on Build GRCh38
rs1043597294 Chr.17: 28547920-28547920 on Build GRCh38
rs1004711099 Chr.17: 28548013-28548013 on Build GRCh38
rs912762781 Chr.17: 28548078-28548078 on Build GRCh38
rs752489401 Chr.17: 28547942-28547942 on Build GRCh38
rs199591380 Chr.17: 28548010-28548010 on Build GRCh38
rs369443469 Chr.17: 28547879-28547879 on Build GRCh38
rs375998799 Chr.17: 28548034-28548034 on Build GRCh38
rs756262246 Chr.17: 28547889-28547889 on Build GRCh38
rs779344508 Chr.17: 28548022-28548022 on Build GRCh38
rs764055633 Chr.17: 28547877-28547877 on Build GRCh38
rs756397372 Chr.17: 28547969-28547969 on Build GRCh38
rs779442575 Chr.17: 28547959-28547959 on Build GRCh38
rs773950656 Chr.17: 28548055-28548055 on Build GRCh38
rs754193171 Chr.17: 28548021-28548021 on Build GRCh38
rs368608427 Chr.17: 28548083-28548083 on Build GRCh38
rs200211619 Chr.17: 28548080-28548080 on Build GRCh38
rs372132951 Chr.17: 28548068-28548068 on Build GRCh38
rs767590360 Chr.17: 28547999-28547999 on Build GRCh38
rs752943228 Chr.17: 28548084-28548084 on Build GRCh38
rs773311477 Chr.17: 28548066-28548066 on Build GRCh38
rs754659345 Chr.17: 28547913-28547913 on Build GRCh38
rs200659406 Chr.17: 28547992-28547992 on Build GRCh38
rs781050872 Chr.17: 28547929-28547929 on Build GRCh38
rs751502943 Chr.17: 28547996-28547996 on Build GRCh38
rs374115959 Chr.17: 28547883-28547883 on Build GRCh38
rs147084613 Chr.17: 28548043-28548043 on Build GRCh38
rs374271322 Chr.17: 28548065-28548065 on Build GRCh38
rs748040933 Chr.17: 28547936-28547936 on Build GRCh38
rs760712949 Chr.17: 28548067-28548067 on Build GRCh38
rs745335874 Chr.17: 28548041-28548041 on Build GRCh38
rs759262279 Chr.17: 28547990-28547990 on Build GRCh38
rs999350588 Chr.17: 28547954-28547954 on Build GRCh38
rs139497685 Chr.17: 28548081-28548081 on Build GRCh38
rs761823478 Chr.17: 28548057-28548057 on Build GRCh38
rs780409623 Chr.17: 28547955-28547955 on Build GRCh38
rs769614921 Chr.17: 28548047-28548047 on Build GRCh38
rs766635958 Chr.17: 28548072-28548072 on Build GRCh38
rs748681105 Chr.17: 28548031-28548031 on Build GRCh38
rs765116657 Chr.17: 28547871-28547871 on Build GRCh38
rs770458721 Chr.17: 28547977-28547977 on Build GRCh38
rs199705690 Chr.17: 28548017-28548017 on Build GRCh38
rs780859514 Chr.17: 28548035-28548035 on Build GRCh38
rs749605669 Chr.17: 28547956-28547956 on Build GRCh38
rs781329096 Chr.17: 28547882-28547882 on Build GRCh38
rs556486188 Chr.17: 28547917-28547917 on Build GRCh38
rs761866874 Chr.17: 28547998-28547998 on Build GRCh38
rs564033763 Chr.17: 28548049-28548049 on Build GRCh38

View Full Product Details