Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67004688-67004961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES4A
Entrez Gene ID
283848
Gene Name
carboxylesterase 4A
Gene Aliases
CES6, CES8
UniGene
Hs.346947
Gene Chromosome Location
Chr.16: 66988589-67009758 on Build GRCh38
Transcripts
NM_001190201.1 NM_001190202.1 NM_001318506.1 NM_173815.6 XM_005255896.4 XM_011523021.2 XM_011523023.2 XM_011523024.2 XM_011523025.2 XM_011523026.2 XM_011523027.2 XM_011523028.2 XM_011523029.2 XM_011523030.2 XM_017023160.1 XM_017023161.1 XM_017023162.1 XM_017023163.1 XM_017023164.1 XM_017023165.1 XM_017023166.1 XM_017023167.1 XM_017023168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4426897 c.1128C>T p.I376I Chr.16: 67004840-67004840 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748541136 Chr.16: 67004814-67004814 on Build GRCh38
rs562910696 Chr.16: 67004717-67004717 on Build GRCh38
rs977598833 Chr.16: 67004787-67004787 on Build GRCh38
rs532152900 Chr.16: 67004907-67004907 on Build GRCh38
rs115953428 Chr.16: 67004822-67004822 on Build GRCh38
rs772170492 Chr.16: 67004806-67004806 on Build GRCh38
rs552166692 Chr.16: 67004917-67004917 on Build GRCh38
rs917846601 Chr.16: 67004895-67004895 on Build GRCh38
rs773799366 Chr.16: 67004919-67004919 on Build GRCh38
rs766441780 Chr.16: 67004815-67004815 on Build GRCh38
rs992066617 Chr.16: 67004862-67004862 on Build GRCh38
rs116885568 Chr.16: 67004780-67004780 on Build GRCh38
rs1031902812 Chr.16: 67004790-67004790 on Build GRCh38
rs1021719397 Chr.16: 67004763-67004763 on Build GRCh38
rs762882232 Chr.16: 67004757-67004757 on Build GRCh38
rs772637227 Chr.16: 67004906-67004906 on Build GRCh38
rs980897174 Chr.16: 67004829-67004829 on Build GRCh38
rs773115768 Chr.16: 67004807-67004807 on Build GRCh38
rs960549176 Chr.16: 67004802-67004802 on Build GRCh38
rs747768546 Chr.16: 67004926-67004927 on Build GRCh38
rs931125102 Chr.16: 67004925-67004925 on Build GRCh38
rs190657959 Chr.16: 67004712-67004712 on Build GRCh38
rs529170797 Chr.16: 67004821-67004821 on Build GRCh38
rs759029199 Chr.16: 67004924-67004925 on Build GRCh38
rs1046862497 Chr.16: 67004936-67004936 on Build GRCh38
rs763955785 Chr.16: 67004891-67004891 on Build GRCh38
rs563163767 Chr.16: 67004857-67004857 on Build GRCh38

View Full Product Details