Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7520509-7520734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF358
Entrez Gene ID
140467
Gene Name
zinc finger protein 358
Gene Aliases
ZFEND
UniGene
Hs.133475
Gene Chromosome Location
Chr.19: 7513880-7521025 on Build GRCh38
Transcripts
NM_018083.4 XM_005272460.3 XM_011527695.1 XM_017026296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7202341 c.1361C>T p.P454L Chr.19: 7520603-7520603 on Build GRCh38
3404774 c.1379G>A p.R460H Chr.19: 7520621-7520621 on Build GRCh38
226815 c.1384C>T p.P462S Chr.19: 7520626-7520626 on Build GRCh38
1271320 c.1312C>T p.L438L Chr.19: 7520554-7520554 on Build GRCh38
2728837 c.1454delC p.S487fs*>82 Chr.19: 7520696-7520696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12977657 Chr.19: 7520637-7520637 on Build GRCh38
rs530318543 Chr.19: 7520626-7520626 on Build GRCh38
rs560758271 Chr.19: 7520664-7520664 on Build GRCh38
rs763706118 Chr.19: 7520601-7520601 on Build GRCh38
rs1039746114 Chr.19: 7520565-7520565 on Build GRCh38
rs768638059 Chr.19: 7520631-7520631 on Build GRCh38
rs749424398 Chr.19: 7520643-7520643 on Build GRCh38
rs532728391 Chr.19: 7520689-7520689 on Build GRCh38
rs745604077 Chr.19: 7520617-7520617 on Build GRCh38
rs756833227 Chr.19: 7520613-7520613 on Build GRCh38
rs540877522 Chr.19: 7520657-7520657 on Build GRCh38
rs979010715 Chr.19: 7520645-7520645 on Build GRCh38
rs12983777 Chr.19: 7520582-7520582 on Build GRCh38
rs12977428 Chr.19: 7520583-7520583 on Build GRCh38
rs866252436 Chr.19: 7520614-7520614 on Build GRCh38
rs367880407 Chr.19: 7520666-7520666 on Build GRCh38
rs1052831400 Chr.19: 7520612-7520612 on Build GRCh38
rs781155547 Chr.19: 7520699-7520699 on Build GRCh38
rs370713422 Chr.19: 7520674-7520674 on Build GRCh38
rs370411153 Chr.19: 7520648-7520648 on Build GRCh38
rs756816223 Chr.19: 7520673-7520673 on Build GRCh38
rs766689955 Chr.19: 7520527-7520527 on Build GRCh38
rs746872690 Chr.19: 7520694-7520694 on Build GRCh38
rs188940050 Chr.19: 7520606-7520606 on Build GRCh38
rs764779004 Chr.19: 7520687-7520698 on Build GRCh38
rs778680031 Chr.19: 7520615-7520615 on Build GRCh38
rs758071669 Chr.19: 7520688-7520688 on Build GRCh38
rs759992139 Chr.19: 7520558-7520558 on Build GRCh38
rs149848212 Chr.19: 7520653-7520653 on Build GRCh38
rs201499923 Chr.19: 7520696-7520696 on Build GRCh38
rs767860377 Chr.19: 7520575-7520575 on Build GRCh38
rs771034646 Chr.19: 7520647-7520647 on Build GRCh38
rs750095655 Chr.19: 7520682-7520682 on Build GRCh38
rs12977668 Chr.19: 7520639-7520639 on Build GRCh38
rs746963840 Chr.19: 7520629-7520629 on Build GRCh38
rs759775922 Chr.19: 7520652-7520652 on Build GRCh38
rs990877013 Chr.19: 7520538-7520538 on Build GRCh38
rs960510184 Chr.19: 7520529-7520529 on Build GRCh38
rs758246034 Chr.19: 7520620-7520620 on Build GRCh38
rs749276767 Chr.19: 7520702-7520702 on Build GRCh38
rs1034784561 Chr.19: 7520703-7520703 on Build GRCh38
rs28374043 Chr.19: 7520600-7520600 on Build GRCh38
rs774546385 Chr.19: 7520540-7520540 on Build GRCh38
rs755614766 Chr.19: 7520597-7520597 on Build GRCh38
rs12977398 Chr.19: 7520560-7520560 on Build GRCh38
rs867225623 Chr.19: 7520618-7520618 on Build GRCh38
rs901243114 Chr.19: 7520581-7520581 on Build GRCh38

View Full Product Details