Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130563150-130563393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A6
Entrez Gene ID
131873
Gene Name
collagen type VI alpha 6
Gene Aliases
-
UniGene
Hs.591282
Gene Chromosome Location
Chr.3: 130516709-130678135 on Build GRCh38
Transcripts
NM_001102608.1 XM_005247121.4 XM_011512423.2 XM_011512424.2 XM_011512425.2 XM_011512426.2 XM_011512428.2 XM_011512431.2 XM_011512432.2 XM_017005711.1 XM_017005712.1 XM_017005713.1 XM_017005714.1 XM_017005715.1 XM_017005716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4480107 c.238C>T p.H80Y Chr.3: 130563241-130563241 on Build GRCh38
1616890 c.306C>G p.G102G Chr.3: 130563309-130563309 on Build GRCh38
4471460 c.171C>T p.L57L Chr.3: 130563174-130563174 on Build GRCh38
1248937 c.273G>A p.L91L Chr.3: 130563276-130563276 on Build GRCh38
1632814 c.381C>A p.D127E Chr.3: 130563384-130563384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765906929 Chr.3: 130563251-130563251 on Build GRCh38
rs904017910 Chr.3: 130563211-130563211 on Build GRCh38
rs771176353 Chr.3: 130563187-130563187 on Build GRCh38
rs200502168 Chr.3: 130563196-130563196 on Build GRCh38
rs748549750 Chr.3: 130563176-130563176 on Build GRCh38
rs747250989 Chr.3: 130563186-130563186 on Build GRCh38
rs772528481 Chr.3: 130563184-130563184 on Build GRCh38
rs1028967299 Chr.3: 130563283-130563283 on Build GRCh38
rs984723306 Chr.3: 130563296-130563296 on Build GRCh38
rs762739997 Chr.3: 130563197-130563197 on Build GRCh38
rs757601806 Chr.3: 130563364-130563364 on Build GRCh38
rs778316628 Chr.3: 130563352-130563352 on Build GRCh38
rs769797783 Chr.3: 130563290-130563290 on Build GRCh38
rs767168248 Chr.3: 130563240-130563240 on Build GRCh38
rs762934753 Chr.3: 130563305-130563305 on Build GRCh38
rs369788872 Chr.3: 130563266-130563266 on Build GRCh38
rs753387960 Chr.3: 130563326-130563326 on Build GRCh38
rs773491093 Chr.3: 130563185-130563185 on Build GRCh38
rs141403454 Chr.3: 130563246-130563246 on Build GRCh38
rs867409186 Chr.3: 130563229-130563229 on Build GRCh38
rs370744388 Chr.3: 130563322-130563322 on Build GRCh38
rs751888547 Chr.3: 130563357-130563357 on Build GRCh38
rs376462458 Chr.3: 130563362-130563362 on Build GRCh38
rs549896105 Chr.3: 130563310-130563310 on Build GRCh38
rs750003075 Chr.3: 130563245-130563245 on Build GRCh38
rs901949993 Chr.3: 130563248-130563248 on Build GRCh38
rs756929368 Chr.3: 130563227-130563227 on Build GRCh38
rs771103950 Chr.3: 130563276-130563276 on Build GRCh38
rs201577074 Chr.3: 130563268-130563268 on Build GRCh38
rs374596678 Chr.3: 130563346-130563346 on Build GRCh38
rs371005676 Chr.3: 130563370-130563370 on Build GRCh38
rs1040186977 Chr.3: 130563180-130563180 on Build GRCh38
rs769989127 Chr.3: 130563369-130563369 on Build GRCh38
rs560384227 Chr.3: 130563189-130563189 on Build GRCh38
rs527993234 Chr.3: 130563212-130563212 on Build GRCh38
rs765180776 Chr.3: 130563195-130563195 on Build GRCh38
rs751357591 Chr.3: 130563219-130563219 on Build GRCh38
rs778062584 Chr.3: 130563267-130563267 on Build GRCh38
rs768634748 Chr.3: 130563309-130563309 on Build GRCh38
rs1016631700 Chr.3: 130563308-130563308 on Build GRCh38
rs760270468 Chr.3: 130563313-130563313 on Build GRCh38
rs776860915 Chr.3: 130563280-130563280 on Build GRCh38
rs567761482 Chr.3: 130563259-130563259 on Build GRCh38
rs775418315 Chr.3: 130563294-130563294 on Build GRCh38
rs748600990 Chr.3: 130563265-130563265 on Build GRCh38
rs746030029 Chr.3: 130563367-130563367 on Build GRCh38
rs745978575 Chr.3: 130563285-130563285 on Build GRCh38
rs961891229 Chr.3: 130563330-130563330 on Build GRCh38
rs750064210 Chr.3: 130563311-130563311 on Build GRCh38
rs781275483 Chr.3: 130563365-130563365 on Build GRCh38
rs751349510 Chr.3: 130563233-130563233 on Build GRCh38

View Full Product Details