Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109667971-109668244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTM2
Entrez Gene ID
2946
Gene Name
glutathione S-transferase mu 2 (muscle)
Gene Aliases
GST4, GSTM, GSTM2-2, GTHMUS
UniGene
Hs.279837
Gene Chromosome Location
Chr.1: 109668022-109683997 on Build GRCh38
Transcripts
NM_000848.3 NM_001142368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2118079 c.32G>A p.R11H Chr.1: 109668147-109668147 on Build GRCh38
2118078 c.32G>A p.R11H Chr.1: 109668147-109668147 on Build GRCh38
6676596 c.34G>A p.G12R Chr.1: 109668149-109668149 on Build GRCh38
2118077 c.32G>A p.R11H Chr.1: 109668147-109668147 on Build GRCh38
6676594 c.34G>A p.G12R Chr.1: 109668149-109668149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377205064 Chr.1: 109668111-109668111 on Build GRCh38
rs766188985 Chr.1: 109668144-109668162 on Build GRCh38
rs116803059 Chr.1: 109668173-109668173 on Build GRCh38
rs989940439 Chr.1: 109668101-109668101 on Build GRCh38
rs752485262 Chr.1: 109668123-109668123 on Build GRCh38
rs546327202 Chr.1: 109668051-109668051 on Build GRCh38
rs999258067 Chr.1: 109668007-109668007 on Build GRCh38
rs1004267076 Chr.1: 109668213-109668213 on Build GRCh38
rs970976478 Chr.1: 109668039-109668039 on Build GRCh38
rs375271866 Chr.1: 109668182-109668182 on Build GRCh38
rs201905887 Chr.1: 109668118-109668118 on Build GRCh38
rs769138224 Chr.1: 109668083-109668083 on Build GRCh38
rs2073483 Chr.1: 109668196-109668196 on Build GRCh38
rs115506091 Chr.1: 109668164-109668164 on Build GRCh38
rs1002486044 Chr.1: 109668224-109668224 on Build GRCh38
rs746457371 Chr.1: 109668066-109668066 on Build GRCh38
rs985049359 Chr.1: 109668044-109668044 on Build GRCh38
rs1024950793 Chr.1: 109668036-109668036 on Build GRCh38
rs912192727 Chr.1: 109668127-109668127 on Build GRCh38
rs773535634 Chr.1: 109668189-109668189 on Build GRCh38
rs367954178 Chr.1: 109668193-109668193 on Build GRCh38
rs772407547 Chr.1: 109668109-109668109 on Build GRCh38
rs774892212 Chr.1: 109668096-109668096 on Build GRCh38
rs906524835 Chr.1: 109668223-109668223 on Build GRCh38
rs766463755 Chr.1: 109668113-109668113 on Build GRCh38
rs758116270 Chr.1: 109668129-109668130 on Build GRCh38
rs530021 Chr.1: 109668158-109668158 on Build GRCh38
rs1018639269 Chr.1: 109668014-109668014 on Build GRCh38
rs751527137 Chr.1: 109668148-109668148 on Build GRCh38
rs575526712 Chr.1: 109668104-109668104 on Build GRCh38
rs625456 Chr.1: 109668049-109668049 on Build GRCh38
rs951966927 Chr.1: 109668043-109668043 on Build GRCh38
rs745482462 Chr.1: 109668153-109668153 on Build GRCh38
rs931120267 Chr.1: 109668165-109668165 on Build GRCh38
rs770477240 Chr.1: 109668076-109668076 on Build GRCh38
rs780933168 Chr.1: 109668079-109668079 on Build GRCh38
rs1033881376 Chr.1: 109668063-109668063 on Build GRCh38
rs907726725 Chr.1: 109668047-109668047 on Build GRCh38
rs561058663 Chr.1: 109668132-109668132 on Build GRCh38
rs754049253 Chr.1: 109668114-109668114 on Build GRCh38
rs147809472 Chr.1: 109668001-109668001 on Build GRCh38
rs1004638967 Chr.1: 109668021-109668021 on Build GRCh38
rs201645642 Chr.1: 109668099-109668099 on Build GRCh38
rs142173575 Chr.1: 109668130-109668130 on Build GRCh38
rs939338203 Chr.1: 109668133-109668133 on Build GRCh38
rs140199111 Chr.1: 109668149-109668149 on Build GRCh38
rs1022436907 Chr.1: 109667995-109667995 on Build GRCh38
rs977501882 Chr.1: 109668120-109668120 on Build GRCh38
rs764986403 Chr.1: 109668122-109668122 on Build GRCh38
rs745572686 Chr.1: 109668080-109668080 on Build GRCh38
rs1046435824 Chr.1: 109668222-109668222 on Build GRCh38
rs751443904 Chr.1: 109668203-109668203 on Build GRCh38
rs554235860 Chr.1: 109668034-109668034 on Build GRCh38
rs995636516 Chr.1: 109667992-109667992 on Build GRCh38
rs776736396 Chr.1: 109668194-109668194 on Build GRCh38
rs752997040 Chr.1: 109668197-109668197 on Build GRCh38
rs887137926 Chr.1: 109668016-109668016 on Build GRCh38
rs764004364 Chr.1: 109668199-109668199 on Build GRCh38

View Full Product Details