Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12646519-12646790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2B1
Entrez Gene ID
4125
Gene Name
mannosidase alpha class 2B member 1
Gene Aliases
LAMAN, MANB
UniGene
Hs.356769
Gene Chromosome Location
Chr.19: 12646508-12666777 on Build GRCh38
Transcripts
NM_000528.3 NM_001173498.1 XM_005259913.1 XM_017026818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3959552 c.2990G>C p.R997P Chr.19: 12646663-12646663 on Build GRCh38
5807594 c.2956C>A p.P986T Chr.19: 12646697-12646697 on Build GRCh38
3822003 c.2933A>C p.H978P Chr.19: 12646720-12646720 on Build GRCh38
6706661 c.2942C>T p.P981L Chr.19: 12646711-12646711 on Build GRCh38
991355 c.2990G>A p.R997H Chr.19: 12646663-12646663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752589383 Chr.19: 12646756-12646756 on Build GRCh38
rs758558306 Chr.19: 12646580-12646580 on Build GRCh38
rs758755293 Chr.19: 12646664-12646664 on Build GRCh38
rs570397849 Chr.19: 12646641-12646641 on Build GRCh38
rs771558548 Chr.19: 12646697-12646697 on Build GRCh38
rs755063947 Chr.19: 12646677-12646677 on Build GRCh38
rs751630577 Chr.19: 12646672-12646672 on Build GRCh38
rs970968464 Chr.19: 12646568-12646568 on Build GRCh38
rs766708959 Chr.19: 12646668-12646668 on Build GRCh38
rs569516380 Chr.19: 12646577-12646577 on Build GRCh38
rs766559062 Chr.19: 12646729-12646729 on Build GRCh38
rs767564979 Chr.19: 12646736-12646736 on Build GRCh38
rs376856949 Chr.19: 12646711-12646711 on Build GRCh38
rs756100832 Chr.19: 12646759-12646759 on Build GRCh38
rs777171557 Chr.19: 12646651-12646651 on Build GRCh38
rs768694116 Chr.19: 12646752-12646754 on Build GRCh38
rs762355600 Chr.19: 12646704-12646704 on Build GRCh38
rs768543157 Chr.19: 12646586-12646586 on Build GRCh38
rs750254194 Chr.19: 12646562-12646562 on Build GRCh38
rs202174515 Chr.19: 12646663-12646663 on Build GRCh38
rs148945108 Chr.19: 12646683-12646683 on Build GRCh38
rs747944082 Chr.19: 12646604-12646604 on Build GRCh38
rs373174565 Chr.19: 12646696-12646696 on Build GRCh38
rs762104115 Chr.19: 12646654-12646654 on Build GRCh38
rs192184706 Chr.19: 12646710-12646710 on Build GRCh38
rs915003108 Chr.19: 12646630-12646630 on Build GRCh38
rs755046385 Chr.19: 12646732-12646732 on Build GRCh38
rs371716069 Chr.19: 12646579-12646579 on Build GRCh38
rs781015466 Chr.19: 12646680-12646680 on Build GRCh38
rs753397171 Chr.19: 12646690-12646690 on Build GRCh38
rs770441236 Chr.19: 12646632-12646632 on Build GRCh38
rs972047517 Chr.19: 12646551-12646551 on Build GRCh38
rs747101669 Chr.19: 12646585-12646585 on Build GRCh38
rs777484621 Chr.19: 12646686-12646686 on Build GRCh38
rs926571642 Chr.19: 12646761-12646761 on Build GRCh38
rs149101136 Chr.19: 12646578-12646578 on Build GRCh38
rs1017209279 Chr.19: 12646726-12646726 on Build GRCh38
rs1019081517 Chr.19: 12646639-12646639 on Build GRCh38
rs866304563 Chr.19: 12646694-12646694 on Build GRCh38
rs200285346 Chr.19: 12646583-12646583 on Build GRCh38
rs369373841 Chr.19: 12646576-12646576 on Build GRCh38
rs899231500 Chr.19: 12646744-12646744 on Build GRCh38
rs777682798 Chr.19: 12646767-12646767 on Build GRCh38
rs749067951 Chr.19: 12646689-12646689 on Build GRCh38
rs749548971 Chr.19: 12646630-12646631 on Build GRCh38
rs763382948 Chr.19: 12646724-12646724 on Build GRCh38
rs764545554 Chr.19: 12646647-12646647 on Build GRCh38
rs772880923 Chr.19: 12646615-12646615 on Build GRCh38
rs573703621 Chr.19: 12646613-12646613 on Build GRCh38
rs1041710709 Chr.19: 12646546-12646546 on Build GRCh38
rs777033630 Chr.19: 12646700-12646700 on Build GRCh38
rs755906293 Chr.19: 12646684-12646684 on Build GRCh38
rs886054228 Chr.19: 12646665-12646665 on Build GRCh38
rs864621991 Chr.19: 12646657-12646657 on Build GRCh38
rs765633432 Chr.19: 12646656-12646656 on Build GRCh38
rs781154896 Chr.19: 12646593-12646593 on Build GRCh38
rs369658144 Chr.19: 12646695-12646695 on Build GRCh38
rs759078247 Chr.19: 12646646-12646646 on Build GRCh38

View Full Product Details