Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78650051-78650302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI44
Entrez Gene ID
10561
Gene Name
interferon induced protein 44
Gene Aliases
MTAP44, TLDC5, p44
UniGene
Hs.82316
Gene Chromosome Location
Chr.1: 78648554-78664078 on Build GRCh38
Transcripts
NM_006417.4 NR_135640.1 NR_135641.1 XM_005270380.3 XM_011540516.2 XM_017000083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934733 c.13A>G p.T5A Chr.1: 78650208-78650208 on Build GRCh38
912408 c.26G>T p.W9L Chr.1: 78650221-78650221 on Build GRCh38
5930564 c.52C>T p.H18Y Chr.1: 78650247-78650247 on Build GRCh38
1296721 c.68G>A p.R23Q Chr.1: 78650263-78650263 on Build GRCh38
3386353 c.24A>G p.T8T Chr.1: 78650219-78650219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958380205 Chr.1: 78650157-78650157 on Build GRCh38
rs780483817 Chr.1: 78650142-78650142 on Build GRCh38
rs928444404 Chr.1: 78650081-78650081 on Build GRCh38
rs148615472 Chr.1: 78650104-78650105 on Build GRCh38
rs966883265 Chr.1: 78650126-78650126 on Build GRCh38
rs1029094690 Chr.1: 78650100-78650100 on Build GRCh38
rs775517843 Chr.1: 78650248-78650248 on Build GRCh38
rs752250083 Chr.1: 78650269-78650269 on Build GRCh38
rs919394011 Chr.1: 78650136-78650136 on Build GRCh38
rs777466431 Chr.1: 78650101-78650101 on Build GRCh38
rs781482789 Chr.1: 78650281-78650281 on Build GRCh38
rs548209725 Chr.1: 78650259-78650259 on Build GRCh38
rs771475743 Chr.1: 78650221-78650221 on Build GRCh38
rs1037491414 Chr.1: 78650091-78650091 on Build GRCh38
rs778499760 Chr.1: 78650206-78650206 on Build GRCh38
rs756152233 Chr.1: 78650166-78650166 on Build GRCh38
rs750339946 Chr.1: 78650158-78650158 on Build GRCh38
rs564036680 Chr.1: 78650239-78650239 on Build GRCh38
rs765099863 Chr.1: 78650265-78650265 on Build GRCh38
rs1008437224 Chr.1: 78650107-78650107 on Build GRCh38
rs766246239 Chr.1: 78650169-78650169 on Build GRCh38
rs781606425 Chr.1: 78650230-78650230 on Build GRCh38
rs929564723 Chr.1: 78650139-78650139 on Build GRCh38
rs758343040 Chr.1: 78650253-78650253 on Build GRCh38
rs375768898 Chr.1: 78650263-78650263 on Build GRCh38
rs111343006 Chr.1: 78650227-78650227 on Build GRCh38
rs763292542 Chr.1: 78650250-78650250 on Build GRCh38
rs990804334 Chr.1: 78650201-78650201 on Build GRCh38
rs142697926 Chr.1: 78650241-78650241 on Build GRCh38
rs566543514 Chr.1: 78650237-78650237 on Build GRCh38
rs148272007 Chr.1: 78650212-78650212 on Build GRCh38
rs754649998 Chr.1: 78650203-78650203 on Build GRCh38
rs2070123 Chr.1: 78650220-78650220 on Build GRCh38
rs758082866 Chr.1: 78650274-78650274 on Build GRCh38
rs374737506 Chr.1: 78650278-78650278 on Build GRCh38
rs772355622 Chr.1: 78650230-78650231 on Build GRCh38
rs367580335 Chr.1: 78650190-78650190 on Build GRCh38
rs147143041 Chr.1: 78650229-78650229 on Build GRCh38
rs544396265 Chr.1: 78650144-78650144 on Build GRCh38
rs1056664705 Chr.1: 78650133-78650133 on Build GRCh38
rs749501287 Chr.1: 78650211-78650211 on Build GRCh38
rs778811769 Chr.1: 78650262-78650262 on Build GRCh38
rs753628573 Chr.1: 78650192-78650192 on Build GRCh38
rs746282455 Chr.1: 78650234-78650234 on Build GRCh38
rs115638974 Chr.1: 78650276-78650276 on Build GRCh38
rs764284663 Chr.1: 78650149-78650149 on Build GRCh38
rs143426079 Chr.1: 78650095-78650095 on Build GRCh38
rs759489899 Chr.1: 78650193-78650194 on Build GRCh38
rs1019737316 Chr.1: 78650117-78650117 on Build GRCh38
rs370707317 Chr.1: 78650242-78650242 on Build GRCh38
rs35977297 Chr.1: 78650161-78650162 on Build GRCh38

View Full Product Details