Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48491387-48491622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMTK3
Entrez Gene ID
114783
Gene Name
lemur tyrosine kinase 3
Gene Aliases
LMR3, PPP1R101, TYKLM3
UniGene
Hs.207426
Gene Chromosome Location
Chr.19: 48485271-48513926 on Build GRCh38
Transcripts
NM_001080434.1 XM_011526411.2 XM_011526412.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4884070 c.4197G>T p.E1399D Chr.19: 48491522-48491522 on Build GRCh38
1395031 c.4218_4219insCC p.E1407fs*>84 Chr.19: 48491500-48491501 on Build GRCh38
6585987 c.4206C>A p.V1402V Chr.19: 48491513-48491513 on Build GRCh38
4698284 c.4185G>A p.T1395T Chr.19: 48491534-48491534 on Build GRCh38
1395029 c.4218_4219insC p.E1407fs*>84 Chr.19: 48491500-48491501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752582954 Chr.19: 48491491-48491491 on Build GRCh38
rs758225154 Chr.19: 48491500-48491501 on Build GRCh38
rs748319479 Chr.19: 48491534-48491534 on Build GRCh38
rs1028391750 Chr.19: 48491468-48491468 on Build GRCh38
rs1018881686 Chr.19: 48491463-48491463 on Build GRCh38
rs753831665 Chr.19: 48491505-48491505 on Build GRCh38
rs977067360 Chr.19: 48491467-48491467 on Build GRCh38
rs771329155 Chr.19: 48491579-48491579 on Build GRCh38
rs759087566 Chr.19: 48491589-48491589 on Build GRCh38
rs1037023601 Chr.19: 48491601-48491601 on Build GRCh38
rs893135802 Chr.19: 48491460-48491460 on Build GRCh38
rs1053166243 Chr.19: 48491458-48491458 on Build GRCh38
rs981607855 Chr.19: 48491585-48491585 on Build GRCh38
rs972387442 Chr.19: 48491578-48491578 on Build GRCh38
rs550348635 Chr.19: 48491596-48491596 on Build GRCh38
rs756006331 Chr.19: 48491502-48491502 on Build GRCh38
rs768903682 Chr.19: 48491509-48491509 on Build GRCh38
rs765544796 Chr.19: 48491446-48491446 on Build GRCh38
rs773791379 Chr.19: 48491450-48491450 on Build GRCh38
rs953354358 Chr.19: 48491439-48491439 on Build GRCh38
rs777732799 Chr.19: 48491501-48491501 on Build GRCh38
rs780233378 Chr.19: 48491507-48491507 on Build GRCh38
rs757133880 Chr.19: 48491506-48491506 on Build GRCh38
rs777666051 Chr.19: 48491503-48491503 on Build GRCh38
rs770157241 Chr.19: 48491551-48491551 on Build GRCh38
rs763584491 Chr.19: 48491567-48491567 on Build GRCh38
rs948896207 Chr.19: 48491515-48491515 on Build GRCh38
rs767257842 Chr.19: 48491490-48491490 on Build GRCh38
rs368871822 Chr.19: 48491483-48491483 on Build GRCh38
rs1011695831 Chr.19: 48491462-48491462 on Build GRCh38
rs372172905 Chr.19: 48491498-48491498 on Build GRCh38
rs186490686 Chr.19: 48491590-48491590 on Build GRCh38
rs953208123 Chr.19: 48491477-48491477 on Build GRCh38
rs140955674 Chr.19: 48491442-48491442 on Build GRCh38
rs527663627 Chr.19: 48491560-48491560 on Build GRCh38
rs781422781 Chr.19: 48491516-48491516 on Build GRCh38
rs763310687 Chr.19: 48491464-48491464 on Build GRCh38
rs773963162 Chr.19: 48491584-48491584 on Build GRCh38
rs552336580 Chr.19: 48491572-48491572 on Build GRCh38
rs747034662 Chr.19: 48491508-48491508 on Build GRCh38
rs111433173 Chr.19: 48491592-48491592 on Build GRCh38
rs989989052 Chr.19: 48491451-48491451 on Build GRCh38
rs759293310 Chr.19: 48491484-48491484 on Build GRCh38

View Full Product Details