Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 48342634-48342865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RB1
Entrez Gene ID
5925
Gene Name
RB transcriptional corepressor 1
Gene Aliases
OSRC, PPP1R130, RB, p105-Rb, pRb, pp110
UniGene
Hs.408528
Gene Chromosome Location
Chr.13: 48303747-48481890 on Build GRCh38
Transcripts
NM_000321.2 XM_011535171.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
317960 c.380+2T>A p.? Chr.13: 48342716-48342716 on Build GRCh38
6851675 c.350T>C p.F117S Chr.13: 48342684-48342684 on Build GRCh38
1367152 c.380G>A p.S127N Chr.13: 48342714-48342714 on Build GRCh38
5659096 c.361C>T p.Q121* Chr.13: 48342695-48342695 on Build GRCh38
4776105 c.380+3A>C p.? Chr.13: 48342717-48342717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777410222 Chr.13: 48342697-48342697 on Build GRCh38
rs1026299929 Chr.13: 48342797-48342797 on Build GRCh38
rs118072861 Chr.13: 48342751-48342751 on Build GRCh38
rs1007840862 Chr.13: 48342840-48342840 on Build GRCh38
rs981839952 Chr.13: 48342789-48342789 on Build GRCh38
rs529855036 Chr.13: 48342786-48342786 on Build GRCh38
rs3092882 Chr.13: 48342731-48342732 on Build GRCh38
rs3092881 Chr.13: 48342726-48342726 on Build GRCh38
rs750711568 Chr.13: 48342742-48342742 on Build GRCh38
rs771961195 Chr.13: 48342679-48342679 on Build GRCh38
rs187110786 Chr.13: 48342724-48342724 on Build GRCh38
rs947950104 Chr.13: 48342709-48342709 on Build GRCh38
rs983885759 Chr.13: 48342693-48342693 on Build GRCh38
rs369249642 Chr.13: 48342828-48342828 on Build GRCh38
rs762953095 Chr.13: 48342720-48342720 on Build GRCh38
rs149800437 Chr.13: 48342701-48342701 on Build GRCh38
rs889382703 Chr.13: 48342810-48342810 on Build GRCh38
rs778932079 Chr.13: 48342670-48342670 on Build GRCh38
rs542686672 Chr.13: 48342819-48342819 on Build GRCh38
rs1052655522 Chr.13: 48342728-48342728 on Build GRCh38
rs559242550 Chr.13: 48342760-48342760 on Build GRCh38
rs951251256 Chr.13: 48342688-48342688 on Build GRCh38
rs139673557 Chr.13: 48342675-48342675 on Build GRCh38
rs767961998 Chr.13: 48342734-48342734 on Build GRCh38
rs869214358 Chr.13: 48342698-48342699 on Build GRCh38
rs769858139 Chr.13: 48342704-48342704 on Build GRCh38
rs775559506 Chr.13: 48342713-48342713 on Build GRCh38
rs576233044 Chr.13: 48342667-48342667 on Build GRCh38
rs984858174 Chr.13: 48342838-48342838 on Build GRCh38
rs774892592 Chr.13: 48342725-48342725 on Build GRCh38
rs748093967 Chr.13: 48342676-48342676 on Build GRCh38
rs520342 Chr.13: 48342759-48342759 on Build GRCh38
rs886042357 Chr.13: 48342710-48342710 on Build GRCh38

View Full Product Details