Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75100513-75100784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC16A5
Entrez Gene ID
9121
Gene Name
solute carrier family 16 member 5
Gene Aliases
MCT5, MCT6
UniGene
Hs.592095
Gene Chromosome Location
Chr.17: 75087727-75106590 on Build GRCh38
Transcripts
NM_001271765.1 NM_004695.3 XM_005257790.1 XM_011525462.1 XM_017025293.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6355241 c.866C>T p.P289L Chr.17: 75100529-75100529 on Build GRCh38
2978517 c.890G>A p.R297Q Chr.17: 75100553-75100553 on Build GRCh38
984267 c.895G>A p.A299T Chr.17: 75100558-75100558 on Build GRCh38
4511107 c.858C>T p.F286F Chr.17: 75100521-75100521 on Build GRCh38
6037997 c.1039G>A p.A347T Chr.17: 75100702-75100702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761272725 Chr.17: 75100552-75100552 on Build GRCh38
rs764413419 Chr.17: 75100745-75100746 on Build GRCh38
rs138922323 Chr.17: 75100738-75100738 on Build GRCh38
rs1000868218 Chr.17: 75100697-75100697 on Build GRCh38
rs116126425 Chr.17: 75100681-75100681 on Build GRCh38
rs776992520 Chr.17: 75100616-75100616 on Build GRCh38
rs75342598 Chr.17: 75100571-75100571 on Build GRCh38
rs761152713 Chr.17: 75100536-75100536 on Build GRCh38
rs141449807 Chr.17: 75100557-75100557 on Build GRCh38
rs763829540 Chr.17: 75100729-75100729 on Build GRCh38
rs752937941 Chr.17: 75100744-75100744 on Build GRCh38
rs748499376 Chr.17: 75100624-75100624 on Build GRCh38
rs115464192 Chr.17: 75100554-75100554 on Build GRCh38
rs189232628 Chr.17: 75100625-75100625 on Build GRCh38
rs141282163 Chr.17: 75100732-75100732 on Build GRCh38
rs749766224 Chr.17: 75100701-75100701 on Build GRCh38
rs776309484 Chr.17: 75100607-75100607 on Build GRCh38
rs773792895 Chr.17: 75100710-75100710 on Build GRCh38
rs769032724 Chr.17: 75100537-75100537 on Build GRCh38
rs149503044 Chr.17: 75100765-75100765 on Build GRCh38
rs748550899 Chr.17: 75100698-75100698 on Build GRCh38
rs765826715 Chr.17: 75100631-75100631 on Build GRCh38
rs1046686603 Chr.17: 75100700-75100700 on Build GRCh38
rs759276639 Chr.17: 75100651-75100651 on Build GRCh38
rs781441141 Chr.17: 75100741-75100741 on Build GRCh38
rs765673955 Chr.17: 75100545-75100545 on Build GRCh38
rs1018021252 Chr.17: 75100763-75100763 on Build GRCh38
rs142646577 Chr.17: 75100574-75100574 on Build GRCh38
rs201506056 Chr.17: 75100566-75100566 on Build GRCh38
rs752605668 Chr.17: 75100657-75100657 on Build GRCh38
rs767266054 Chr.17: 75100656-75100656 on Build GRCh38
rs138345813 Chr.17: 75100731-75100731 on Build GRCh38
rs746000787 Chr.17: 75100578-75100578 on Build GRCh38
rs778013551 Chr.17: 75100759-75100759 on Build GRCh38
rs539147742 Chr.17: 75100674-75100674 on Build GRCh38
rs764144547 Chr.17: 75100671-75100671 on Build GRCh38
rs775263317 Chr.17: 75100727-75100727 on Build GRCh38
rs755019385 Chr.17: 75100739-75100739 on Build GRCh38
rs759113830 Chr.17: 75100722-75100722 on Build GRCh38
rs377663371 Chr.17: 75100699-75100699 on Build GRCh38
rs759058500 Chr.17: 75100745-75100745 on Build GRCh38
rs556632401 Chr.17: 75100553-75100553 on Build GRCh38
rs777566991 Chr.17: 75100576-75100576 on Build GRCh38
rs760358306 Chr.17: 75100728-75100728 on Build GRCh38
rs568762538 Chr.17: 75100640-75100640 on Build GRCh38
rs370678017 Chr.17: 75100573-75100573 on Build GRCh38
rs755946691 Chr.17: 75100660-75100660 on Build GRCh38
rs762311764 Chr.17: 75100544-75100544 on Build GRCh38
rs767427505 Chr.17: 75100569-75100569 on Build GRCh38
rs747260583 Chr.17: 75100595-75100595 on Build GRCh38
rs375463804 Chr.17: 75100628-75100628 on Build GRCh38
rs767323165 Chr.17: 75100726-75100726 on Build GRCh38
rs775544421 Chr.17: 75100535-75100535 on Build GRCh38
rs761060135 Chr.17: 75100574-75100575 on Build GRCh38
rs140468386 Chr.17: 75100691-75100691 on Build GRCh38
rs758387023 Chr.17: 75100582-75100582 on Build GRCh38
rs753744110 Chr.17: 75100673-75100673 on Build GRCh38
rs771413277 Chr.17: 75100702-75100702 on Build GRCh38
rs747029613 Chr.17: 75100687-75100687 on Build GRCh38
rs973521965 Chr.17: 75100668-75100668 on Build GRCh38
rs147815125 Chr.17: 75100556-75100556 on Build GRCh38
rs766092741 Chr.17: 75100555-75100555 on Build GRCh38
rs145115821 Chr.17: 75100737-75100737 on Build GRCh38
rs1002825516 Chr.17: 75100591-75100591 on Build GRCh38
rs151010040 Chr.17: 75100632-75100632 on Build GRCh38

View Full Product Details