Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 75424585-75424844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC15
Entrez Gene ID
158866
Gene Name
zinc finger DHHC-type containing 15
Gene Aliases
MRX91
UniGene
Hs.253211
Gene Chromosome Location
Chr.X: 75368427-75523626 on Build GRCh38
Transcripts
NM_001146256.1 NM_144969.2 XM_006724624.3 XM_017029296.1 XM_017029297.1 XM_017029298.1

SNP Detail

SNP ID Chromosome Location
rs866052920 Chr.X: 75424781-75424781 on Build GRCh38
rs770220728 Chr.X: 75424779-75424779 on Build GRCh38
rs754900931 Chr.X: 75424676-75424676 on Build GRCh38
rs763511839 Chr.X: 75424792-75424792 on Build GRCh38
rs771215114 Chr.X: 75424755-75424755 on Build GRCh38
rs760209616 Chr.X: 75424820-75424820 on Build GRCh38
rs764347182 Chr.X: 75424795-75424795 on Build GRCh38
rs763313778 Chr.X: 75424608-75424608 on Build GRCh38
rs777671319 Chr.X: 75424711-75424711 on Build GRCh38
rs371039365 Chr.X: 75424620-75424620 on Build GRCh38
rs764511227 Chr.X: 75424613-75424613 on Build GRCh38
rs377715509 Chr.X: 75424765-75424765 on Build GRCh38
rs908373477 Chr.X: 75424659-75424659 on Build GRCh38
rs1028202520 Chr.X: 75424621-75424622 on Build GRCh38
rs760058155 Chr.X: 75424707-75424707 on Build GRCh38
rs778995331 Chr.X: 75424680-75424680 on Build GRCh38
rs753423990 Chr.X: 75424670-75424670 on Build GRCh38
rs376637014 Chr.X: 75424612-75424612 on Build GRCh38
rs747167251 Chr.X: 75424743-75424743 on Build GRCh38
rs745960509 Chr.X: 75424766-75424766 on Build GRCh38
rs139863988 Chr.X: 75424656-75424656 on Build GRCh38
rs952589824 Chr.X: 75424627-75424627 on Build GRCh38
rs755848143 Chr.X: 75424653-75424653 on Build GRCh38
rs750033101 Chr.X: 75424623-75424623 on Build GRCh38
rs371358966 Chr.X: 75424797-75424797 on Build GRCh38
rs200759801 Chr.X: 75424686-75424686 on Build GRCh38
rs758251200 Chr.X: 75424696-75424696 on Build GRCh38

View Full Product Details