Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 100334810-100335083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WARS
Entrez Gene ID
7453
Gene Name
tryptophanyl-tRNA synthetase
Gene Aliases
GAMMA-2, IFI53, IFP53
UniGene
Hs.497599
Gene Chromosome Location
Chr.14: 100333788-100376343 on Build GRCh38
Transcripts
NM_004184.3 NM_173701.1 NM_213645.1 NM_213646.1 XM_005268044.3 XM_006720249.3 XM_011537133.2 XM_011537135.2 XM_011537136.2 XM_017021627.1 XM_017021628.1 XM_017021629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6821183 c.1259A>G p.Y420C Chr.14: 100335032-100335032 on Build GRCh38
4049071 c.1282G>A p.G428S Chr.14: 100335009-100335009 on Build GRCh38
3361065 c.1311T>G p.V437V Chr.14: 100334980-100334980 on Build GRCh38
6139366 c.1354G>A p.V452I Chr.14: 100334937-100334937 on Build GRCh38
6051797 c.1404C>T p.F468F Chr.14: 100334887-100334887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775527812 Chr.14: 100335025-100335025 on Build GRCh38
rs755527132 Chr.14: 100335041-100335041 on Build GRCh38
rs765377416 Chr.14: 100334933-100334933 on Build GRCh38
rs377079808 Chr.14: 100335017-100335017 on Build GRCh38
rs764382529 Chr.14: 100335027-100335027 on Build GRCh38
rs778768863 Chr.14: 100335048-100335048 on Build GRCh38
rs775279931 Chr.14: 100334887-100334887 on Build GRCh38
rs753004827 Chr.14: 100334935-100334935 on Build GRCh38
rs763195088 Chr.14: 100335026-100335026 on Build GRCh38
rs371089804 Chr.14: 100335031-100335031 on Build GRCh38
rs375615815 Chr.14: 100335063-100335063 on Build GRCh38
rs79878944 Chr.14: 100334893-100334893 on Build GRCh38
rs746989249 Chr.14: 100335053-100335053 on Build GRCh38
rs748720876 Chr.14: 100335015-100335015 on Build GRCh38
rs766502615 Chr.14: 100334925-100334925 on Build GRCh38
rs772125168 Chr.14: 100335004-100335004 on Build GRCh38
rs753471733 Chr.14: 100335045-100335045 on Build GRCh38
rs745465377 Chr.14: 100334879-100334879 on Build GRCh38
rs756709800 Chr.14: 100334863-100334863 on Build GRCh38
rs187196177 Chr.14: 100334895-100334895 on Build GRCh38
rs768529026 Chr.14: 100334965-100334965 on Build GRCh38
rs201020165 Chr.14: 100334980-100334980 on Build GRCh38
rs952755077 Chr.14: 100334916-100334916 on Build GRCh38
rs762599454 Chr.14: 100335036-100335037 on Build GRCh38
rs759020649 Chr.14: 100335051-100335052 on Build GRCh38
rs908634296 Chr.14: 100334977-100334977 on Build GRCh38
rs72713916 Chr.14: 100334994-100334994 on Build GRCh38
rs759722365 Chr.14: 100334932-100334932 on Build GRCh38
rs751027202 Chr.14: 100335043-100335044 on Build GRCh38
rs779643935 Chr.14: 100335043-100335043 on Build GRCh38
rs536195792 Chr.14: 100335058-100335058 on Build GRCh38
rs372639869 Chr.14: 100334945-100334945 on Build GRCh38
rs774379129 Chr.14: 100334901-100334901 on Build GRCh38
rs530395314 Chr.14: 100335010-100335010 on Build GRCh38
rs747956658 Chr.14: 100335051-100335051 on Build GRCh38
rs778223361 Chr.14: 100334841-100334841 on Build GRCh38
rs368813835 Chr.14: 100334869-100334869 on Build GRCh38
rs140956308 Chr.14: 100334906-100334906 on Build GRCh38
rs758740628 Chr.14: 100334936-100334936 on Build GRCh38
rs773270850 Chr.14: 100335009-100335009 on Build GRCh38
rs776589951 Chr.14: 100334926-100334926 on Build GRCh38
rs977484422 Chr.14: 100334934-100334934 on Build GRCh38
rs79907075 Chr.14: 100334886-100334886 on Build GRCh38
rs768048960 Chr.14: 100335055-100335058 on Build GRCh38
rs769723507 Chr.14: 100335055-100335055 on Build GRCh38
rs749896038 Chr.14: 100334949-100334949 on Build GRCh38
rs2234533 Chr.14: 100335047-100335047 on Build GRCh38
rs771946654 Chr.14: 100335052-100335052 on Build GRCh38
rs112867811 Chr.14: 100335018-100335018 on Build GRCh38
rs139914390 Chr.14: 100334964-100334964 on Build GRCh38
rs747565490 Chr.14: 100334859-100334859 on Build GRCh38
rs1036030640 Chr.14: 100335059-100335059 on Build GRCh38
rs751453486 Chr.14: 100334948-100334948 on Build GRCh38
rs778852654 Chr.14: 100334971-100334971 on Build GRCh38
rs976993442 Chr.14: 100334996-100334996 on Build GRCh38
rs751907243 Chr.14: 100335029-100335029 on Build GRCh38
rs550196955 Chr.14: 100335036-100335036 on Build GRCh38
rs764466208 Chr.14: 100334937-100334937 on Build GRCh38
rs755689812 Chr.14: 100334960-100334960 on Build GRCh38
rs765836940 Chr.14: 100335040-100335041 on Build GRCh38

View Full Product Details