Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178964659-178964926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF454
Entrez Gene ID
285676
Gene Name
zinc finger protein 454
Gene Aliases
-
UniGene
Hs.259441
Gene Chromosome Location
Chr.5: 178941191-178966433 on Build GRCh38
Transcripts
NM_001178089.2 NM_001178090.2 NM_001323306.1 NM_182594.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3342294 c.477G>T p.M159I Chr.5: 178964881-178964881 on Build GRCh38
3342293 c.466G>A p.G156R Chr.5: 178964870-178964870 on Build GRCh38
7135732 c.365G>A p.C122Y Chr.5: 178964769-178964769 on Build GRCh38
316812 c.417A>C p.R139R Chr.5: 178964821-178964821 on Build GRCh38
5778599 c.522C>A p.S174R Chr.5: 178964926-178964926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756394064 Chr.5: 178964848-178964848 on Build GRCh38
rs6867221 Chr.5: 178964859-178964859 on Build GRCh38
rs184153007 Chr.5: 178964757-178964757 on Build GRCh38
rs769421704 Chr.5: 178964781-178964781 on Build GRCh38
rs866730849 Chr.5: 178964891-178964891 on Build GRCh38
rs759005394 Chr.5: 178964801-178964801 on Build GRCh38
rs777490761 Chr.5: 178964855-178964855 on Build GRCh38
rs749187567 Chr.5: 178964791-178964791 on Build GRCh38
rs202137968 Chr.5: 178964694-178964694 on Build GRCh38
rs1046268717 Chr.5: 178964834-178964834 on Build GRCh38
rs770872824 Chr.5: 178964796-178964796 on Build GRCh38
rs188558963 Chr.5: 178964832-178964832 on Build GRCh38
rs746601386 Chr.5: 178964884-178964884 on Build GRCh38
rs12719860 Chr.5: 178964901-178964901 on Build GRCh38
rs780232283 Chr.5: 178964729-178964729 on Build GRCh38
rs747965316 Chr.5: 178964759-178964759 on Build GRCh38
rs781348651 Chr.5: 178964842-178964842 on Build GRCh38
rs775061413 Chr.5: 178964813-178964813 on Build GRCh38
rs201056410 Chr.5: 178964819-178964819 on Build GRCh38
rs751739255 Chr.5: 178964746-178964746 on Build GRCh38
rs762148959 Chr.5: 178964684-178964684 on Build GRCh38
rs771424891 Chr.5: 178964872-178964872 on Build GRCh38
rs758978924 Chr.5: 178964726-178964726 on Build GRCh38
rs139248906 Chr.5: 178964826-178964826 on Build GRCh38
rs775069119 Chr.5: 178964879-178964879 on Build GRCh38
rs367751228 Chr.5: 178964784-178964784 on Build GRCh38
rs568431052 Chr.5: 178964877-178964877 on Build GRCh38
rs369566427 Chr.5: 178964836-178964836 on Build GRCh38
rs773826555 Chr.5: 178964799-178964799 on Build GRCh38
rs778885859 Chr.5: 178964869-178964869 on Build GRCh38
rs143152818 Chr.5: 178964858-178964858 on Build GRCh38
rs200467241 Chr.5: 178964902-178964902 on Build GRCh38
rs773682052 Chr.5: 178964896-178964896 on Build GRCh38
rs765453823 Chr.5: 178964820-178964820 on Build GRCh38
rs768472563 Chr.5: 178964888-178964888 on Build GRCh38
rs763532667 Chr.5: 178964825-178964825 on Build GRCh38
rs146210796 Chr.5: 178964696-178964696 on Build GRCh38
rs752798303 Chr.5: 178964844-178964844 on Build GRCh38
rs371714816 Chr.5: 178964807-178964807 on Build GRCh38
rs755073147 Chr.5: 178964840-178964840 on Build GRCh38
rs745368254 Chr.5: 178964870-178964870 on Build GRCh38

View Full Product Details